Key Documents
GE18-1129-59
EPS power supply adapters
2 mm male/4 mm female
About This Item
UNSPSC Code:
41121815
NACRES:
NB.22
feature
wetted part: no
packaging
pkg of 1 ea
manufacturer/tradename
Cytiva 18112959
Features and Benefits
  • Choose adapters for connecting 4 mm or 2 mm leads to 2 mm or 4 mm output jacks, respectively.
  • This adapter is approved for 5NANA V relative protection ground.
  • Use Adapter Set 2mm M/4mm F for connecting sheathed 2 mm male plug instrument leads to 4 mm sheathed output jacks.
  • Use Adapter Set 4mm M/2mm F for connecting sheathed 4 mm male plug instrument leads to 2 mm high-voltage output jacks.
Storage Class Code
13 - Non Combustible Solids
Choose from one of the most recent versions:
Certificates of Analysis (COA)