Skip to Content
Merck
All Photos(1)

Key Documents

GE17-5132-02

Glutathione Sepharose 4 Fast Flow

Cytiva 17-5132-02, pack of 100 mL

Synonym(s):

Sepharose 4 Fast Flow

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106500
NACRES:
NA.56

ligand

glutathione and 10-carbon linker arm

packaging

pack of 100 mL

manufacturer/tradename

Cytiva 17-5132-02

matrix

highly cross-linked 4% agarose

average diameter

90 μm

cleaning

3-12

working range

3-12

capacity

>10 mg/mL binding capacity

Related Categories

General description

Binding of GST to glutathione is flow-rate dependent and lower flow rates often increase the binding capacity. This is important to consider during sample loading and elution.
Glutathione Sepharose 4 Fast Flow is an affinity medium for fast, one-step purification of Glutathione S-Transferase (GST) tagged proteins. Provides good flow properties, excellent for scale-up.

Glutathione Sepharose 4 Fast Flow is used for the purification of GST-tagged proteins from bacterial lysates and other S-tranferases or glutathione-dependent proteins. Mild elution conditions preserve protein antigenicity and function. Sepharose 4 Fast Flow provides good flow properties and is excellent for scaling up.

Features and Benefits

  • Fast, one-step purification of GST-tagged proteins
  • Mild elution conditions preserving protein antigenicity and function
  • For scale-up due to high flow properties
  • Glutathione coupled to Sepharose 4 Fast Flow via a 10-carbon spacer arm

Storage and Stability

Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.
Store at 4 to 30 °C (20% Ethanol)

Analysis Note

To view the Certificate of Analysis for this product, please visit www.cytiva.com.

Legal Information

Sepharose is a trademark of Cytiva

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Storage Class Code

3 - Flammable liquids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

This page describes general considerations for purification of GST-tagged proteins using Cytiva products.

GST-tagged protein yield varies based on protein, host cell, and culture conditions, influencing production.

GST-tagged protein yield varies based on protein, host cell, and culture conditions, influencing production.

GST-tagged protein yield varies based on protein, host cell, and culture conditions, influencing production.

See All

Protocols

This page shows how to purify GST-tagged proteins using Glutathione Sepharose from Cytiva.

GSTrap™ affinity columns are specially designed 1 ml and 5 ml HiTrap® columns packed with Glutathione Sepharose High Performance, Glutathione Sepharose 4 Fast Flow, or Glutathione Sepharose 4B media.

This page how to remove GST tags by enzymatic cleavage using Cytiva products.

Affinity chromatography tags like GST fusion proteins facilitate separations in various purification products from Cytiva.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service