Skip to Content
Merck
All Photos(1)

Key Documents

GE17-0554-02

Agarose NA

Cytiva 17-0554-02, pack of 100 g

Synonym(s):

Agarose gel

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105323
NACRES:
NA.25
Pricing and availability is not currently available.

form

solid

feature

wetted part: no

packaging

pack of 100 g

manufacturer/tradename

Cytiva 17-0554-02

color

White

Related Categories

General description

Agarose NA has been prepared to give the optimal combination of these properties: Reliable separation of nucleic acids; low inhibition of enzyme activity on recovered material; and high gel strength.

Application

Agarose with Low electroendosmosis (EEO) is recommended for most DNA/RNA applications. If separated nucleic acids are to be used as substrates for enzymes (e.g., ligase and restriction endonucleases), a special highly purified Agarose such as Agarose NA should be used.

For preparative work, an Agarose with Low gelling temperature should be used. Agarose Prep has Low melting and gelling points.

Features and Benefits

  • Highly suitable for DNA/RNA applications.

Storage and Stability

Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.

Analysis Note

To view the Certificate of Analysis for this product, please visit www.cytiva.com.

Storage Class Code

13 - Non Combustible Solids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Protocols

This page describes typical rehydration solutions for use in gel electrophoresis with Cytiva products.

Related Content

This page shows how to apply equilibrated Immobiline DryStrip gels

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service