Skip to Content
Merck
All Photos(2)

Key Documents

GBL612107

Grace Bio-Labs HybriWell sealing system, SecureSeal adhesive chamber

chamber volume 50 μL, L × W × thickness 9.8 mm × 20 mm × 0.25 mm, port diam. 1.5 mm

Synonym(s):

chamber slides, chambered coverglass, cover slides, hybridization chambers, incubation chambers

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122600
NACRES:
NB.22

material

colorless
polycarbonate

sterility

non-sterile

feature

wells 6

packaging

pack of 100 ea

manufacturer/tradename

Grace Bio-Labs 612107

L × W × thickness

9.8 mm × 20 mm × 0.25 mm

chamber volume

50 μL

port diam.

1.5 mm

wells

3

Looking for similar products? Visit Product Comparison Guide

General description

Grace Bio-Labs HybriWell sealing system seals securely to a microscope slide surface in seconds to accommodate waterbath hybridizations, such as protein or nucleic acid hybridization assays.

Features:
  • Encloses a single or multiple specimens in a small reagent volume and eliminate evaporation
  • Access ports in the chamber surface allow for the addition or removal of reactants
  • Ports are easily sealed using adhesive seal tabs provided with each order
  • Sealed chambers are watertight and ideally suited for waterbath incubations
  • RNase free, hydrophobic surfaces will not trap or bind probes
  • Disposable chamber removes cleanly and easily even after heating
  • Each package includes illustrated instructions for use, applicator and 200 adhesive port seals

Legal Information

HybriWell is a trademark of Grace Bio-Labs, Inc.
SecureSeal is a trademark of Grace Bio-Labs, Inc.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service