Skip to Content
Merck
All Photos(1)

Key Documents

G9420

Sigma-Aldrich

GNF-2

≥98% (HPLC), solid

Synonym(s):

3-[6-[[4-(Trifluoromethoxy)phenyl]amino]-4-pyrimidinyl]benzamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H13F3N4O2
CAS Number:
Molecular Weight:
374.32
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98% (HPLC)

form

solid

color

off-white

solubility

H2O: <2 mg/mL
DMSO: ≥5 mg/mL

storage temp.

2-8°C

SMILES string

NC(=O)c1cccc(c1)-c2cc(Nc3ccc(OC(F)(F)F)cc3)ncn2

InChI

1S/C18H13F3N4O2/c19-18(20,21)27-14-6-4-13(5-7-14)25-16-9-15(23-10-24-16)11-2-1-3-12(8-11)17(22)26/h1-10H,(H2,22,26)(H,23,24,25)

InChI key

WEVYNIUIFUYDGI-UHFFFAOYSA-N

Biochem/physiol Actions

GNF-2 belongs to a new class of Bcr-abl inhibitors. GNF-2 appears to bind to the myristoyl binding pocket, an allosteric site distant from the active site, stabilizing the inactive form of the kinase. It inhibits Bcr-abl phosphorylation with an IC50 of 267 nM, but does not inhibit a panel of 63 other kinases, including native c-Abl, and shows complete lack of toxicity towards cells not expressing Bcr-Abl. GNF-2 shows great potential for a new class of inhibitor to study Bcr-abl activity and to treat resistant Chronic myelogenous leukemia (CML), which is caused the Bcr-Abl oncoprotein.

Features and Benefits

This compound is a featured product for Kinase Phosphatase Biology research. Click here to discover more featured Kinase Phosphatase Biology products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.
This compound is featured on the Abl page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Melissanne de Wispelaere et al.
Cell chemical biology, 25(8), 1006-1016 (2018-06-26)
Viral envelope proteins are required for productive viral entry and initiation of infection. Although the humoral immune system provides ample evidence for targeting envelope proteins as an antiviral strategy, there are few pharmacological interventions that have this mode of action.
Francisco J Adrián et al.
Nature chemical biology, 2(2), 95-102 (2006-01-18)
Chronic myelogenous leukemia (CML) is a myeloproliferative disorder characterized at the molecular level by the expression of Bcr-abl, a 210-kDa fusion protein with deregulated tyrosine kinase activity. Encouraged by the clinical validation of Bcr-abl as the target for the treatment
Inhibitors of Bcr-abl... breaking new ground again.
Jeffrey F Ohren et al.
Nature chemical biology, 2(2), 63-64 (2006-01-20)
So-Youn Kim et al.
Cell death and differentiation, 26(3), 502-515 (2018-07-11)
Platinum-based chemotherapies can result in ovarian insufficiency by reducing the ovarian reserve, a reduction believed to result from apoptosis of immature oocytes via activation/phosphorylation of TAp63α by multiple kinases including CHEK2, CK1, and ABL1. Here we demonstrate that cisplatin (CDDP)
Haibin Tong et al.
Journal of cellular biochemistry, 119(3), 2806-2817 (2017-10-24)
The excessive recruitment and improper activation of polymorphonuclear neutrophils (PMNs) often induces serious injury of host tissues, leading to inflammatory disorders. Therefore, to understand the molecular mechanism on neutrophil recruitment possesses essential pathological and physiological importance. In this study, we

Articles

Discover Bioactive Small Molecules for Kinase Phosphatase Biology

Discover Bioactive Small Molecules for Kinase Phosphatase Biology

Discover Bioactive Small Molecules for Kinase Phosphatase Biology

Discover Bioactive Small Molecules for Kinase Phosphatase Biology

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service