Key Documents
F7677
Sigma-Aldrich
Nunc® EasYFlasks, Nunclon® Δ Surface
Nunclon® Δ surface treatment, 175 cm2 angled neck, filter cap, 30/cs
Synonym(s):
cell culture flask, tissue culture flask, tissue culture treated flask
About This Item
UNSPSC Code:
41122104
NACRES:
NB.22
Pricing and availability is not currently available.
material
canted neck transparent polystyrene flask
screw top blue high-density polyethylene cap (filter)
sterility
sterile; γ-irradiated
feature
with cap
packaging
case of 30 ea (6 packs x 5 flasks)
manufacturer/tradename
Nunc 159910
flask working volume
55 mL
surface area
175 cm2
fitting
33 mm cap
binding type
Nunclon Delta surface
General description
Nunc® EasYFlasks, Nunclon® Δ Surface
  • Nunclon® Δ certified: a proprietary cell culture surface treatment that offers maximum adhesion for a broad range of cell types. The ergonomic design with angled neck and wide opening allows easy access to the growth surface by pipettes or cell scrapers.
  • Ergonomic design with a 1/3 turn to open, close or vent
  • Low profile to maximize incubator space
  • Volume graduations printed and molded on both sides
  • Slightly angled side walls for complete viewing under a microscope
  • Sterilized by gamma irradiation and certified to be nonpyrogenic
  • Nunclon Delta Certification, including testing on four different cell types, provided in each case
  • Vent/close caps feature a “Y” mark that allows visual verification of vent position.
  • Vent position easily distinguished even when flasks are stacked in incubators.
  • Filter membrane provides effective barrier against micro-organisms.
  • Well-defined, hydrophobic filter membrane offers continuous venting with consistent gas exchange and no lot-to-lot variation.
  • Chemically robust membrane meets USP class VI standards and is produced without animal-derived substances.
Legal Information
EasYFlasks is a trademark of Thermo Fisher Scientific or its subsidiaries
Nunc is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Nunclon is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Visit the Document Library
Customers Also Viewed
Slide 1 of 1
1 of 1
Characterization of colony-forming cells in adult human articular cartilage.
Ozbey O, Sahin Z, Acar N, et al.
Acta Histochemica, 116(5), 763-770 (2014)