Documents
EP5391070034
Eppendorf(R) Plate Adapter HeatSealer S100
for 96 and 384 well plates, high profile
About This Item
UNSPSC Code:
41121815
NACRES:
NB.15
feature
for 96 and 384 well plates
high profile
manufacturer/tradename
Eppendorf® 5391070034 (Global)
Features and Benefits
Hermetic Sealing, of multiwell plates
Eliminates evaporation in PCR
Seals 96- and 384-well plates
Integrated thermostat prevents overheating
Improved mechanics for even easier Sealing,
PCR Plate Adapter included
Legal Information
Eppendorf is a registered trademark of Eppendorf AG