Skip to Content
Merck
All Photos(1)

Key Documents

EP3116000015

Sigma-Aldrich

Eppendorf® Pipette Holder System

Pipette Carousel 2, for 6 Eppendorf Research™, Eppendorf Research™ plus, Reference™, Reference™ 2 or Biomaster™

Synonym(s):

pipette storage, pipettor holder

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121500
NACRES:
NB.02
Pricing and availability is not currently available.

material

plastic

description

For use with Eppendorf Research™, Eppendorf Research™ plus, Eppendorf Reference™, Eppendorf Reference™ 2, Biomaster™

feature

Holds 6 x pipetors
for 6 Eppendorf Research, Eppendorf Research plus, Reference, Reference 2 or Biomaster

packaging

pack of 1 ea

manufacturer/tradename

Eppendorf® 3116000015

Looking for similar products? Visit Product Comparison Guide

General description

Keep all your Eppendorf pipettes and Multipettes well within reach. The Pipette Carousel 2 for manual pipettes and the Charger Carousel 2 for electronic devices off er more fl exibility and more capacity than ever before. The Eppendorf Pipette Carousel 2 carries up to six manual single or multi-channel Eppendorf pipettes. Single pipette holders can be mounted to the carousel in two orientations turned by 180 degrees to each other to store all current and most predecessor manual pipettes. This includes Eppendorf Research, Research plus, Reference, Reference 2 and Biomaster®. The Charger Carousel 2 offers 50% more capacity than before and holds up to six electronic Eppendorf Xplorer or Xplorer plus pipettes. When used with optional charger shells the carousel can carry up to six Eppendorf Multipette E3/E3x or Multipette stream/Xstream. In addition to electronic devices, the Charger Carousel 2 is also perfect for manual Eppendorf pipettes when used with optional pipette holders
The Eppendorf pipette holder system provides safe and convenient storage. As all pipettes are positioned upright, their nose cone does not touch the lab bench reducing the risk of damage and cross contamination. The carousels provide easy access to the pipette of your choice and have a robust design with a small footprint. They can hold up to six liquid handling instruments. The stands carry one electronic pipette or Multipette. Carousels and stands are available with and without charging functionality. The wall mount devices store one manual or electronic pipette or Multipette.

Application

Liquid handling

Features and Benefits

More capacity
  • Charger Carousel 2 for up to six electronic devices
  • Carousels carry both manual and electronic devices
More convenience
  • Interchangeable pipette holders, no tools required
  • Cord wrap functionality for unused power cord More user safety
  • Magnetic power connector for fast and easy connection
  • Large rubber feet protect carousels from liquids spilled on bench top
New Eppendorf Pipette Holder System stores all current manual and electronic Eppendorf pipettes (including the Multipette) and most of their predecessors
Charger carousel with intelligent charging electronics and increased capacity for up to six instruments
Rotatable holder with completely new design for all manual pipettes including Eppendorf Research®, Eppendorf Research® plus, Reference®, Reference® 2 and Biomaster®
Carousels and stands with interchangeable pipette holders and charger shells, no tools required
Large rubber feet protect carousels and stands from liquids spilled on bench top
Pictograms on all charger shells and pipette holders for clear assignment in the lab

Legal Information

Biomaster is a registered trademark of Eppendorf AG
Eppendorf is a registered trademark of Eppendorf AG
Eppendorf Research is a registered trademark of Eppendorf AG
Reference is a registered trademark of Eppendorf AG

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service