Skip to Content
Merck
All Photos(1)

Key Documents

EP0030129504

Eppendorf twin.tec® PCR Plates 96

Semi-Skirted, LoBind, PCR clean, working volume 250 μL, colorless, pack of 25

Synonym(s):

cell culture plate, multi well plates, multiple well plates

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106300
NACRES:
NB.22
material:
colorless
size:
96 well
well maximum volume:
250 μm
working volume:
250 μL
sterility:
non-sterile
feature:
skirt
LoBind
PCR clean
Semi-Skirted
Pricing and availability is not currently available.

material

colorless

well size

96

sterility

non-sterile

feature

skirt
LoBind
PCR clean
Semi-Skirted

packaging

pack of 25

manufacturer/tradename

Eppendorf® 0030129504 (Global)

technique(s)

PCR: suitable

size

96 well

well maximum volume

250 μm

working volume

250 μL

Looking for similar products? Visit Product Comparison Guide

General description

  • Eppendorf® LoBind twin.tec® PCR plates are available as a one-piece design combining a polycarbonate frame and polypropylene wells.
  • LoBind material ensures minimized sample-to–surface binding for optimal product recovery.
  • Thin-walled wells ensure optimal heat transfer.
  • OptiTrack® matrix for high contrast labeling of the alphanumeric grid for accurate sample identification and to minimize pipetting errors.
  • Raised well rims facilitate effective sealing and prevent cross-contamination.
  • Free from detectable human DNA, DNase, RNase, and PCR inhibitors.
Choose Eppendorf twin.tec PCR plates to achieve reliable PCR results every time. twin.tec plates offer purity, stability, different colors and other simple, intelligent features that make your everyday work easier. The stable polycarbonate frame is well suited for automated and high-throughput processes, and the different colors allow you to quickly organize the individual steps of your workflow.

Application

  • PCR with low template concentration
  • Real-time PCR with low sample concentration
  • Low volume PCR/qPCR
  • NGS DNA library construction
PCR

Features and Benefits

  • Maximum recovery rates for improved assay results
  • Available with barcode (upon request)
  • Ideal for quantitative real-time PCR as well
  • Exceptionally solid and torque-resistant polycarbonate frame
  • OptiTrack® matrix for faster sample identification and fewer pipetting errors
  • Raised well rims for effective sealing, also reduces risk of cross-contamination
  • Batch-tested and certified free of DNA, DNAase, RNase and PCR inhibitors (PCR clean)

Legal Information

Eppendorf is a registered trademark of Eppendorf AG
OptiTrack is a registered trademark of Eppendorf AG
twin.tec is a registered trademark of Eppendorf AG

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service