EP0030118430
Eppendorf® Conical Tubes

sterile, cap (snapTech ), endotoxin free, Biopur, DNase free, Biopur, capacity 25 mL, amber, pkg of 150 ea (6 bags x 25 tubes)
About This Item
material
amber
cap (snapTech )
conical bottom
polyethylene cap
polypropylene
sterility
sterile
feature
DNase free Biopur
PCR clean Biopur
RNase free Biopur
endotoxin free Biopur
graduation: no
packaging
pkg of 150 ea (6 bags x 25 tubes)
manufacturer/tradename
Eppendorf® 0030118430
parameter
17,000 x g rpm speed
capacity
25 mL
color
, light protection
suitability
suitable for PCR
Looking for similar products? Visit Product Comparison Guide
General description
Please note the Instructions For Use for details ((URL: https://www.eppendorf.com/manuals)[www.eppendorf.com/manuals]): "Eppendorf Conical Tubes 25 mL, snap cap" and "Eppendorf Conical Tubes 15, 25 and 50 mL"
Features and Benefits
- High degree of transparency for full control of insertion depth of your pipette tip
- Effective protection of UV light-sensitive samples
- Centrifugation stability up to 17,000 x g1) for both tube variants - SnapTec cap and screw cap
- Tightness of the lid is ensured over a broad temperature range
- Less storage space needed due to ~20 % lower height compared to 50 mL conical tubes
- Matching accessories for centrifugation, mixing and heating, sample preparation and storage ensure straightforward integration into existing laboratory environment
- Premium USP class 6 raw material and no use of slip agents, plasticizers, and biocides during manufacturing ensure highest sample integrity and optimal sample and pellet visibility
- Single-handed opening and closing through SnapTec cap version for fast liquid extraction and addition
- Reduced height gives easy sample access even with low-volume pipettes and tips – reduced risk of cross-contamination between pipette and tube
- The 25 mL conical tube with SnapTec cap is autoclavable at 121 °C in 20 min.
Please note the Instructions For Use for details ((URL: https://www.eppendorf.com/manuals)[www.eppendorf.com/manuals]): "Eppendorf Conical Tubes 25 mL, snap cap" and "Eppendorf Conical Tubes 15, 25 and 50 mL"
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service