Skip to Content
Merck
All Photos(2)

Key Documents

EP0030113578

Eppendorf twin.tec® PCR Plates 384

skirt, PCR clean, working volume 40 μL, colorless (with 2D SafeCode), pkg of 25 ea

Synonym(s):

Plates

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122102
NACRES:
NB.15
material:
colorless (with 2D SafeCode)
polypropylene
round wells
size:
384 well
sterility:
non-sterile
feature:
PCR clean
lid: no
skirt

material

colorless (with 2D SafeCode)
polypropylene
round wells

sterility

non-sterile

feature

PCR clean
lid: no
skirt

packaging

pkg of 25 ea

manufacturer/tradename

Eppendorf® 0030113578

size

384 well

working volume

40 μL

suitability

suitable for PCR

Looking for similar products? Visit Product Comparison Guide

General description

Eppendorf twin.tec PCR Plate 384, skirted, 40 µL, PCR clean, colorless with 2D SafeCode (DataMatrix/ECC200) and human readable code, 25 plates
The Eppendorf twin.tec PCR plate combines extremely thin-walled polypropylene wells for optimal heat transfer to the sample, with robust polycarbonate frames for high rigidity. With raised well rims, twin.tec plates enable highly effective sealing to reduce the risk of cross-contamination.

Features and Benefits

  • 3-level coding for safe sample and vessel identification
  • Codes on three sides of plates for flexible reading
  • ECC 200 error correction function enables safe code reading with up to 30 % damaged codes
  • Additional, ID-specific information on plates available via Eppendorf DataPort for documentation purposes
  • Direct import into eLabInventory and eLabJournal software for convenient handling
  • Import to other database systems possible
  • Manufactured from high-quality polypropylene for highest sample purity

Legal Information

Eppendorf is a registered trademark of Eppendorf AG
twin.tec is a registered trademark of Eppendorf AG

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service