Skip to Content
Merck
All Photos(1)

Documents

EHU131171

Sigma-Aldrich

MISSION® esiRNA

targeting human USP7

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

Quality Level

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

TACGTGACTTGCTCCCAGTTATGTGTGACAGAGCAGGATTTATTCAAGATACTAGCCTTATCCTCTATGAGGAAGTTAAACCGAATTTAACAGAGAGAATTCAGGACTATGACGTGTCTCTTGATAAAGCCCTTGATGAACTAATGGATGGTGACATCATAGTATTTCAGAAGGATGACCCTGAAAATGATAACAGTGAATTACCCACCGCAAAGGAGTATTTCCGAGATCTCTACCACCGCGTTGATGTCATTTTCTGTGATAAAACAATCCCTAATGATCCTGGATTTGTGGTTACGTTATCAAATAGAATGAATTATTTTCAGGTTGCAAAGACAGTTGCACAGAGGCTCAACACAGATCCAATGTTGCTGCAGTTTTTCAAGTCTCAAGGTTATAGGGATGGCCCAGGT

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yufei Wang et al.
EMBO reports, 20(7), e47563-e47563 (2019-07-04)
Monoubiquitination of histone H2B on lysine 120 (H2Bub1) is an epigenetic mark generally associated with transcriptional activation, yet the global functions of H2Bub1 remain poorly understood. Ferroptosis is a form of non-apoptotic cell death characterized by the iron-dependent overproduction of
Xiaomeng Dai et al.
Theranostics, 10(20), 9332-9347 (2020-08-18)
Background: Tumor associated macrophages (TAMs) have strong plasticity and if reprogrammed, can clear tumor cells and regulate the adaptive immune system for cancer immunotherapy. Deubiquitinating enzymes (DUBs), which can remove ubiquitin (Ub) from Ub-modified substrates, have been associated with oncogenic
Peiyi Xie et al.
Molecular and cellular endocrinology, 518, 111037-111037 (2020-09-24)
Ubiquitin-specific protease 7 (USP7/HAUSP) is known to regulate multiple cellular phenomena, including cell cycle progression and proliferation, and is involved in binding and stabilizing specific target proteins through deubiquitylation. However, the detailed role of USP7 in papillary thyroid carcinoma (PTC)
Giovanna Carrà et al.
Oncotarget, 8(22), 35508-35522 (2017-04-19)
Chronic Lymphocytic Leukemia (CLL) is a lymphoproliferative disorder with either indolent or aggressive clinical course. Current treatment regiments have significantly improved the overall outcomes even if higher risk subgroups - those harboring TP53 mutations or deletions of the short arm
Eduardo de la Vega et al.
The FEBS journal, 287(21), 4659-4677 (2020-03-03)
Satellite cells (SCs) are myogenic progenitors responsible for skeletal muscle regeneration and maintenance. Upon activation, SCs enter a phase of robust proliferation followed by terminal differentiation. Underlying this myogenic progression, the sequential expression of muscle regulatory transcription factors (MRFs) and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service