Skip to Content
Merck
All Photos(1)

Key Documents

EHU054741

Sigma-Aldrich

MISSION® esiRNA

targeting human PRKD1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

AGCTTCTGAAGGGGCTTTTCAGGCAGGGCTTGCAGTGCAAAGATTGCAGATTCAACTGCCATAAACGTTGTGCACCGAAAGTACCAAACAACTGCCTTGGCGAAGTGACCATTAATGGAGATTTGCTTAGCCCTGGGGCAGAGTCTGATGTGGTCATGGAAGAAGGGAGTGATGACAATGATAGTGAAAGGAACAGTGGGCTCATGGATGATATGGAAGAAGCAATGGTCCAAGATGCAGAGATGGCAATGGCAGAGTGCCAGAACGACAGTGGCGAGATGCAAGATCCAGACCCAGACCACGAGGACGCCAACAGAACCATCAGTCCATCAACAAGCAACAATATCCCACTCATGAGGGTAGTGCAGTCTGTCAAACACACGAAGAGGAAAAGCAGCAC

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kimberly A Coughlan et al.
The Journal of biological chemistry, 291(11), 5664-5675 (2016-01-23)
AMP-activated protein kinase (AMPK) is an energy-sensing enzyme whose activity is inhibited in settings of insulin resistance. Exposure to a high glucose concentration has recently been shown to increase phosphorylation of AMPK at Ser(485/491) of its α1/α2 subunit; however, the
Qi Zhou et al.
Archives of toxicology, 93(6), 1639-1648 (2019-04-26)
2,3',4,4',5-Pentachlorobiphenyl (PCB118) has been shown to cause thyroidal ultrastructure lesions, but the underlying mechanism remains elusive. This study aimed to elucidate the mechanism by which PCB118 induces the abnormalities of the thyrocytes. Wistar rats were injected intraperitoneally with PCB118 (0
Di Zhao et al.
International journal of biological sciences, 13(3), 276-285 (2017-04-04)
Growing evidence shows that protein kinase D (PKD) plays an important role in the development of pressure overload-induced cardiac hypertrophy. However, the mechanisms involved are not clear. This study tested our hypothesis that PKD might mediate cardiac hypertrophy by negatively
Jia Wang et al.
American journal of physiology. Cell physiology, 310(7), C542-C557 (2016-01-08)
Given the fundamental role of β-catenin signaling in intestinal epithelial cell proliferation and the growth-promoting function of protein kinase D1 (PKD1) in these cells, we hypothesized that PKDs mediate cross talk with β-catenin signaling. The results presented here provide several
Jonathan Baker et al.
PloS one, 13(4), e0195864-e0195864 (2018-04-14)
Many catabolic stimuli, including interleukin-1 (IL-1) in combination with oncostatin M (OSM), promote cartilage breakdown via the induction of collagen-degrading collagenases such as matrix metalloproteinase 1 (MMP1) and MMP13 in human articular chondrocytes. Indeed, joint diseases with an inflammatory component

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service