Skip to Content
Merck
All Photos(1)

Key Documents

CLS3844

Corning® 384 well plate, clear, poly-D-Lysine, with lid

clear polystyrene, flat bottom, non-sterile, lid, case of 100

Synonym(s):

Corning® 384 well microplate

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121800
NACRES:
NB.13
material:
clear polystyrene
flat bottom
polystyrene plate
size:
384 wells
Pricing and availability is not currently available.

material

clear polystyrene
flat bottom
polystyrene plate

sterility

non-sterile

feature

lid

packaging

case of 100

manufacturer/tradename

Corning 3844

availability

available only in South Korea

size

384 wells

well volume

95 μL

well working volume

20-80 μL

color

clear

suitability

suitable for (cell cuture)

Looking for similar products? Visit Product Comparison Guide

Legal Information

Corning is a registered trademark of Corning, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service