Key Documents
CLS3762
Corning® 384 well microplate, low flange
black polystyrene, flat bottom, non-sterile, lid: no, pack of 25
Synonym(s):
384 multiwell plates, 384 well low volume plates, 384 well microplates, 384 well microtiter plates, 384 well plates, 384 well tissue culture treated plates
About This Item
UNSPSC Code:
41121800
NACRES:
NB.24
material:
black polystyrene
clear bottom
flat bottom
polystyrene
size:
384 wells
Pricing and availability is not currently available.
material
black polystyrene
clear bottom
flat bottom
polystyrene
sterility
non-sterile
feature
lid: no
skirt
packaging
case of 100 ea
pack of 25
manufacturer/tradename
Corning 3762
maximum volume
112 μL
size
384 wells
working volume
80 μL
suitability
suitable for (cell cuture)
binding type
non-treated surface
General description
  • Corning® 384 well microplate, low flange black polystyrene, flat bottom, non-sterile, with no lid conforms to standard microplate footprint and dimensions.
  • Untreated (or medium binding) polystyrene surface is hydrophobic.
  • Applicable for fluorescent assays using either top or bottom detection plate readers.
  • Recommended working volume of up to 20 to 80 μL.
  • Binding capacity of approximately 100 to 200ng IgG/cm².
Legal Information
Corning is a registered trademark of Corning, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)