Skip to Content
Merck
All Photos(1)

Key Documents

PE/CA-PJ15

96121230, human tongue, squamous cell carcinoma. Epithelial-like morphology.

Synonym(s):

Human PE/CA-PJ15, PE/CA-PJ15 Cell Line

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

Product Name

PE/CA-PJ15, 96121230

biological source

human mouth (epithelial cells)

growth mode

Adherent

karyotype

Not specified

morphology

Epithelial-like

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Human oral squamous cell carcinoma

Cell Line Description

The cell line PE/CA-PJ15 was established from tongue tissue of a 45 year old male patient with oral squamous cell carcinoma. The cells have been shown to have epithelial differentiated characteristics expressing laminin and cytokeratins. Involucrin has been detected in some cells and cell groups.

Application

Study of squamous cell carcinoma

Culture Medium

IDMEM + 2mM Glutamine + 10% Foetal Bovine Serum (FBS).

Subculture Routine

Split sub-confluent cultures (70-80%) 1:2 to 1:8 using 0.25% trypsin or trypsin/EDTA; 5% CO2; 37°C. Note: cells may take up to 7 days to become confluent, a medium change during this period may be required.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service