Skip to Content
Merck
All Photos(1)

Key Documents

PNT2 Cell Line human

95012613, human prostate, Epithelial

Synonym(s):

PNT-2 Cells

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
biological source:
human prostate
growth mode:
Adherent
karyotype:
Not specified
morphology:
Epithelial
products:
Not specified
receptors:
Not specified

Product Name

PNT2 Cell Line human, Normal prostate epithelium immortalized with SV40., 95012613

biological source

human prostate

growth mode

Adherent

karyotype

Not specified

morphology

Epithelial

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Human prostate normal, immortalised with SV40

Cell Line Description

Established by immortalisation of normal adult prostatic epithelial cells by transfection with a plasmid containing SV40 genome with a defective replication origin. The primary culture was obtained from a prostate of a 33 year old male at post mortem. PNT2 cells contain the SV40 genome and express large T protein. They possess a well differentiated morphology with the expression of cytokeratin 8,18 and 19 with the latter being a feature of differentiated luminal cells of the glandular prostate. Cytokeratin 14, a marker of epithelial basal cells, is not expressed. The cells are non-tumourigenic in nude mice.

Application

Biology of prostate cells, model of differentiated prostate epithelial cells.
PNT2 cell line has been used:
  • to assess the effects of Verapamil, an L-type calcium channel blocker on autophagy
  • to assess the cytotoxicity of aqueous dilutions of a universal self-priming dental adhesive (DA)
  • to study the expression of soluble adenylyl cyclase (sAC) in the cells

DNA Profile

STR-PCR Data: Amelogenin: X,Y
CSF1PO: 11,13
D13S317: 8,13
D16S539: 9
D5S818: 12,13
D7S820: 11
THO1: 6,9.3
TPOX: 8,9
vWA: 17

Culture Medium

RPMI 1640 + 2mM Glutamine + 6 -10% Foetal Bovine Serum (FBS).

Subculture Routine

Split sub-confluent cultures (70-80%) 1:3 to 1:10 i.e. seeding at 2-4x10,000 cells/cm2 using 0.25% trypsin or trypsin/EDTA; 5% CO2; 37°C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service