Skip to Content
Merck
All Photos(1)

Key Documents

COLO 684

87061203, human uterus, Round & Refractile

Synonym(s):

COLO684 Cells

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

Product Name

COLO 684, 87061203

biological source

human uterus

growth mode

Suspension

karyotype

Not specified

morphology

Round & Refractile

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Human uterus adenocarcinoma

Cell Line Description

Derived from an adenocarcinoma of the uterus from a 63 year old patient. COLO 685 (Sigma Catalogue number 87061206) was established from the same patient as confirmed by STR profilling.

Application

Tumourigenicity studies

Culture Medium

RPMI 1640 + 2mM Glutamine + 10% Foetal Bovine Serum (FBS).

Subculture Routine

Maintain cultures between 3-9x100,000 cells/ml; 5% CO2; 37°C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service