Skip to Content
Merck
All Photos(1)

Key Documents

A7r5 Cell Line from rat

86050803, rat muscle, Fibroblast

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
Pricing and availability is not currently available.

Product Name

A7r5 Cell Line from rat, 86050803

biological source

rat muscle

description

Rat BDIX smooth muscle embryonic aorta

growth mode

Adherent

karyotype

Not specified

morphology

Fibroblast

products

Muscle-type isoenzymes

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Rat BDIX smooth muscle embryonic aorta

Cell Line Description

Flat ribbon-like cells differentiating to parallel arrays of spindle shaped cells. Derived from the thoracic aorta of an embryonic BDIX rat. As cells achieve the stationary phase there is an increase in myokinase and creatine phosphokinase (CPK) activity. After cessation of cell division a muscle type CPK isoenzyme is synthesised.

Application

A7r5 Cell Line from rat has been used to study the effect of angiotensin II on vascular oxidative stress.[1] It has been used in PLA2 inhibitory study to understand the role of PLA2 on formation of lipid droplets.[2]
Biochemical and biophysical studies

Culture Medium

DMEM + 2mM Glutamine + 10% Foetal Bovine Serum (FBS).

Subculture Routine

Split sub-confluent cultures (70-80%) 1:2 to 1:3 i.e. seeding at 3-5x10,000 cells/cm2 using 0.25% trypsin; 5% CO2; 37°C. After resuscitation cells may take up to 7 days before a monolayer forms and growth can be very slow in general. Cells also grow in clumps.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service