Skip to Content
Merck
All Photos(1)

Documents

P3X63Ag8

85011401, mouse skin, Lymphoblast

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

product name

P3X63Ag8, 85011401

biological source

mouse skin

growth mode

Suspension

karyotype

Aneuploid

morphology

Lymphoblast

products

Immunoglobulin G (IgG1) (kappa)

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Mouse myeloma

Cell Line Description

Derived from the P3X27 cell line, itself a derivative of mineral oil-induced plasmacytoma MOPC-21 in BALB/c mice. Resistant to 20μg/ml azaguanine, does not grow in selective medium. HGPRT neg., dies in HAT. Doubling time 16-26 h.

Application

Fusion partner

Culture Medium

DMEM or RPMI 1640 + 2mM Glutamine + 10% Foetal Bovine Serum (FBS).

Subculture Routine

Maintain cultures between 5x10,000 to 3x100,000 cells/ml; 5% CO2; 37°C. Cells will achieve maximum cell density of 5x100,000 cells/ml.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Selection of the right fusion partner cell line is critical to obtain desired attributes of the hybridoma. Explore hybridoma fusion partners cell lines.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service