Skip to Content
Merck
All Photos(1)

Key Documents

C1103

Sigma-Aldrich

Anti-Calcium Channel (α1 Subunit), Pan antibody produced in rabbit

affinity isolated antibody, lyophilized powder

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

lyophilized powder

species reactivity

mouse, rat

technique(s)

immunohistochemistry: suitable using rat heart sections (The epitope can be masked by other subunits in some Ca2+ channels.)
western blot: 1:200 using rat brain membranes

UniProt accession no.

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

Specificity

The epitope recognized by this antibody is masked by α2 and δ chains; therefore, the antibody binds only after subunit dissociation using Triton X-100 or other detergents.

Immunogen

synthetic peptide corresponding to amino acids 1506-1524 of the α1S subunit of rat skeletal muscle voltage-gated calcium channel (VGCC, CP15) (Accession P22002) with additional N-terminal cysteine, conjugated to MBS-KLH.

Application

Anti-Calcium Channel (α1 Subunit), Pan antibody produced in rabbit is suitable for immunohistochemistry (using rat heart sections) and western blotting at a dilution of 1:200 using rat brain membranes.
Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Immunocytochemistry (1 paper)

Biochem/physiol Actions

Calcium channel, voltage-dependent, L type, α1C subunit (Cav1.2) is a protein encoded by the CACNA1C gene in humans and belongs to the family of voltage-gated calcium channels. It serves as the key transducers of cell surface membrane potential changes into local intracellular calcium transients that initiate different physiological events. CACNA1C is a subunit of L-type voltage-dependent calcium channel. It is mapped to chromosome 12p13.3 and is implicated as a susceptibility gene for schizophrenia. CACNA1C is selectively expressed in human TH2 cells and blockage of Cav1.2 channel activation in TH2 cells may represent new strategies to treat allergic diseases in human subjects. Its increased activity is implicated in the pathogenesis of dementia and Alzheimer′s disease (AD).

Physical form

Lyophilized from phosphate buffered saline, pH 7.4, containing 1% bovine serum albumin, 5% sucrose and 0.025% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Verena Burtscher et al.
Biochimica et biophysica acta, 1838(8), 2053-2065 (2014-05-07)
Defective retinal synaptic transmission in patients affected with congenital stationary night blindness type 2 (CSNB2) can result from different dysfunction phenotypes in Cav1.4 L-type calcium channels. Here we investigated two prototypical Cav1.4 variants from either end of the functional spectrum.
Ju Hwan Kim et al.
General physiology and biophysics, 38(5), 379-388 (2019-08-15)
With the rapidly increasing use of mobile phones and their close-contact usage to the brain, there are some concerns about the possible neuronal effects induced by exposure to excessive electromagnetic radiation. Exposure to a radiofrequency electromagnetic field (RF-EMF) of 835
Fanfan Zheng et al.
Schizophrenia research, 152(1), 105-110 (2013-12-21)
CACNA1C (12p13.3) has been implicated as a susceptibility gene for schizophrenia by several replicated genome wide association studies. While these results have been consistent among studies in European populations, the findings in East Asian populations have varied. To test whether
Johannes Voelker et al.
Cells, 10(10) (2021-10-24)
Causal therapies for the auditory-pathway and inner-ear diseases are still not yet available for clinical application. Regenerative medicine approaches are discussed and examined as possible therapy options. Neural stem cells could play a role in the regeneration of the auditory
William A Catterall et al.
Pharmacological reviews, 57(4), 411-425 (2005-12-31)
The family of voltage-gated calcium channels serves as the key transducers of cell surface membrane potential changes into local intracellular calcium transients that initiate many different physiological events. There are 10 members of the voltage-gated calcium channel family that have

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service