Skip to Content
Merck
All Photos(1)

Key Documents

AXYTR222Y

Pipette reference tips

size 200 μL, pkg of 20x1000tips/cs, yellow

Synonym(s):

pipet tips

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121600
NACRES:
NB.02

material

yellow

sterility

non-sterile

feature

DNA free
RNase free
barrier: no
endotoxin free
filter: no
graduation

manufacturer/tradename

Axygen Inc TR-222-Y
Corning

packaging

pkg of 20x1000tips/cs

size

200 μL

color

yellow

Looking for similar products? Visit Product Comparison Guide

Other Notes

Browse all pipette tips

Legal Information

Corning is a registered trademark of Corning, Inc.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service