Skip to Content
Merck
All Photos(4)

Key Documents

AV48205

Sigma-Aldrich

Anti-GOT1 antibody produced in rabbit

IgG fraction of antiserum

Synonym(s):

Anti-GIG18, Anti-Glutamic-oxaloacetic transaminase 1, soluble (aspartate aminotransferase 1)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

46 kDa

species reactivity

horse, human, rabbit, goat, mouse, rat, guinea pig, bovine

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... GOT1(2805)

General description

Glutamic-oxaloacetic transaminase 1, soluble (GOT1) is an enzyme that is involved in the metabolism of amino acids. It is also involved in tricarboxylic acid and area cycles. Studies have reported that GOT1 plays a role in vesicle formation from the ER. GOT1 has been xenografted to nude mice for radiotherapy studies.
Rabbit Anti-GOT1 antibody recognizes human, mouse, rat, bovine, pig, and chicken GOT1.

Immunogen

Synthetic peptide directed towards the N terminal region of human GOT1

Application

Rabbit Anti-GOT1 antibody is suitable for western blot applications at a concentration of 5μg/ml.

Biochem/physiol Actions

Glutamic-oxaloacetic transaminase is a pyridoxal phosphate-dependent enzyme which exists in cytoplasmic and mitochondrial forms, GOT1 and GOT2, respectively. GOT plays a role in amino acid metabolism and the urea and tricarboxylic acid cycles. The two enzymes are homodimeric and show close homology.Glutamic-oxaloacetic transaminase is a pyridoxal phosphate-dependent enzyme which exists in cytoplasmic and mitochondrial forms, GOT1 and GOT2, respectively. GOT plays a role in amino acid metabolism and the urea and tricarboxylic acid cycles. The two enzymes are homodimeric and show close homology. Publication Note: This RefSeq record includes a subset of the publications that are available for this gene. Please see the Entrez Gene record to access additional publications.

Sequence

Synthetic peptide located within the following region: MAPPSVFAEVPQAQPVLVFKLTADFREDPDPRKVNLGVGAYRTDDCHPWV

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Andrés Lorente-Rodríguez et al.
Journal of cell science, 122(Pt 10), 1540-1550 (2009-04-23)
Yip1p belongs to a conserved family of membrane-spanning proteins that are involved in intracellular trafficking. Studies have shown that Yip1p forms a heteromeric integral membrane complex, is required for biogenesis of ER-derived COPII vesicles, and can interact with Rab GTPases.
Ola Nilsson et al.
Annals of the New York Academy of Sciences, 1014, 275-279 (2004-05-22)
Malignant carcinoid tumors express high numbers of somatostatin receptors. Radiation therapy using labeled somatostatin analogs is a novel treatment modality for these tumors. We have analyzed the biokinetics and therapeutic effect of radiolabeled somatostatin analog on a human midgut carcinoid
Jia Liu et al.
BMC cardiovascular disorders, 22(1), 492-492 (2022-11-21)
To investigate the role of circNFIB in the alleviation of myocardial fibrosis by endogenous sulfur dioxide (SO2). We stimulated cultured neonatal rat cardiac fibroblasts with transforming growth factor-β1 (TGF-β1) and developed an in vitro myocardial fibrosis model. Lentivirus vectors containing
Jaime Abrego et al.
Cancer discovery, 12(10), 2414-2433 (2022-07-28)
Despite significant recent advances in precision medicine, pancreatic ductal adenocarcinoma (PDAC) remains near uniformly lethal. Although immune-modulatory therapies hold promise to meaningfully improve outcomes for patients with PDAC, the development of such therapies requires an improved understanding of the immune
Tatsunori Suzuki et al.
Cancer gene therapy (2021-04-10)
Mutational activation of the KRAS gene occurs in almost all pancreatic ductal adenocarcinoma (PDAC) and is the earliest molecular event in their carcinogenesis. Evidence has accumulated of the metabolic reprogramming in PDAC, such as amino acid homeostasis and autophagic flux.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service