Skip to Content
Merck
All Photos(1)

Key Documents

AV44247

Sigma-Aldrich

Anti-MICA antibody produced in rabbit

IgG fraction of antiserum

Synonym(s):

Anti-DAQB-48K1.7, Anti-MGC111087, Anti-MHC class I polypeptide-related sequence A, Anti-PERB11.1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
clone:
polyclonal
application:
WB
species reactivity:
human
technique(s):
western blot: suitable
citations:
4

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

42 kDa

species reactivity

human

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... MICA(4276)

Immunogen

Synthetic peptide directed towards the middle region of human MICA

Application

Anti-MICA antibody produced in rabbit is suitable for western blotting at a concentration of 2.5μg/ml.

Biochem/physiol Actions

MHC Class I polypeptide-related sequences A and B (MICA and MICB) are surface glycoproteins expressed constitutively on the enterocytes. They act as ligands for the natural killer group 2, member D (NKG2D) immunoreceptor activation. MICA binds NKG2D receptor and activates the CD8+T cells. Tissue damage or increased expression of MICA results in auto-antibodies detected in early-onset systemic lupus erythematosus and celiac disease.

Sequence

Synthetic peptide located within the following region: LRRTVPPMVNVTRSEASEGNITVTCRASGFYPWNITLSWRQDGVSLSHDT

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Antonio López-Vázquez et al.
BMC medicine, 12, 34-34 (2014-02-26)
Overexpression of autologous proteins can lead to the formation of autoantibodies and autoimmune diseases. MHC class I polypeptide-related sequence A (MICA) is highly expressed in the enterocytes of patients with celiac disease, which arises in response to gluten. The aim
Zhenpeng Dai et al.
The Journal of experimental medicine, 206(4), 793-805 (2009-03-18)
The NKG2D receptor stimulates natural killer cell and T cell responses upon engagement of ligands associated with malignancies and certain autoimmune diseases. However, conditions of persistent NKG2D ligand expression can lead to immunosuppression. In cancer patients, tumor expression and shedding
S Bahram et al.
Proceedings of the National Academy of Sciences of the United States of America, 91(14), 6259-6263 (1994-07-05)
Major histocompatibility complex (MHC) class I genes typically encode polymorphic peptide-binding chains which are ubiquitously expressed and mediate the recognition of intracellular antigens by cytotoxic T cells. They constitute diverse gene families in different species and include the numerous so-called
Natasja Nielsen et al.
Immunology, 142(4), 581-593 (2014-03-29)
Rheumatoid arthritis (RA) is an autoimmune disease characterized by chronic inflammation and synovial hyperplasia leading to progressive joint destruction. Fibroblast-like synoviocytes (FLS) are central components of the aggressive, tumour-like synovial structure termed pannus, which invades the joint space and cartilage.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service