Skip to Content
Merck
All Photos(1)

Documents

93052608

COLO 699 N

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

biological source

human lymph

Quality Level

form

liquid

growth mode

Adherent

karyotype

Not specified

morphology

Epithelial

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

General description

COLO 699 N cells, a non-small cell lung cancer cell line is useful in subcutaneous xenografts studies to mimic in vivo model of human lung cancer. It finds applications in stable isotope-based proteome analysis.

Cell Line Origin

Human lung cancer

Cell Line Description

Derived from the pleural fluid of a 57 year old female with lung cancer. Please note: This cell line was found to be indistinguishable from cell lines RPMI 7932 & COLO 738 by STR PCR DNA profiling. At ECACC the STR PCR profile of all stocks of this cell line match the profile of the original source material received at ECACC. However, it is not clear if the cell lines COLO 699N, RPMI 7932 & COLO 738 were originally derived from the same individual as the profiles suggest. The cell lines COLO 699N, RPMI 7932 & COLO 738 were received from the same depositor. If we are able to obtain clarification of the relationship between these cell lines this information will be updated.

Application

COLO 699 N has been used in cytotoxicity studies with Tegaran and 13-methyltetradecanoic acid (13-MTD).
Tumourigenicity studies

DNA Profile

STR-PCR Data: Amelogenin: X
CSF1PO: 10
D13S317: 11
D16S539: 12
D5S818: 11,12
D7S820: 11,13
THO1: 9,9.3
TPOX: 8
vWA: 17

Culture Medium

RPMI 1640 + 2mM Glutamine + 10% Foetal Bovine Serum (FBS).

Subculture Routine

Split sub-confluent cultures (70-80%) 1:3 to 1:6 i.e. seeding at 2-4x10,000 cells/cm2using 0.25% trypsin/EDTA; 5% CO2; 37°C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Disclaimer

RESEARCH USE ONLY. This product is regulated in France when intended to be used for scientific purposes, including for import and export activities (Article L 1211-1 paragraph 2 of the Public Health Code). The purchaser (i.e. enduser) is required to obtain an import authorization from the France Ministry of Research referred in the Article L1245-5-1 II. of Public Health Code. By ordering this product, you are confirming that you have obtained the proper import authorization.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A S Boehle et al.
The Annals of thoracic surgery, 69(4), 1010-1015 (2000-05-09)
Overall prognosis in human lung cancer is still poor. A highly reproducible, easy to perform in vivo model, which closely resembles the clinical features of advanced human lung cancer, is required for the evaluation of novel therapies. Tumor cells, originated
Kathrin Grundner-Culemann et al.
Journal of proteomics, 130, 1-10 (2015-09-13)
Non-small cell lung cancer (NSCLC) cell lines are widely used model systems to study molecular aspects of lung cancer. Comparative and in-depth proteome expression data across many NSCLC cell lines has not been generated yet, but would be of utility
H G Morse et al.
Cancer genetics and cytogenetics, 69(2), 108-112 (1993-09-01)
Melanoma cell lines initiated from metastases excised at the same time from multiple sites in a patient reflect a single clone origin. The similarities of the karyotypes of these lines depend upon the site of excision, the selective survival in
Panagiotis Parsonidis et al.
PloS one, 15(10), e0240969-e0240969 (2020-10-22)
The aim of this study is to evaluate the potential health effects of Tegaran Formula ZhenHua, a nutritional supplement used mainly by cancer patients. Its active ingredients and cytotoxicity was assessed with analytical methods and viability assays, respectively. The analytical

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service