Skip to Content
Merck
All Photos(1)

Key Documents

5090

Sigma-Aldrich

CDH18 human

≥90% (SDS-PAGE), recombinant, expressed in E. coli, liquid

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.75

Product Name

CDH18 human, recombinant, expressed in E. coli, 0.5 mg protein/mL

biological source

human

recombinant

expressed in E. coli

description

0.1 mg of recombinant human CDH18 in 20 mM Tris-HCl buffer, containing NaCl, KCl, EDTA, L-arginine, DTT and glycerol.

sterility

Filtered sterilized solution

Assay

≥90% (SDS-PAGE)

form

liquid

packaging

pkg of 100 μg

concentration

0.5 mg protein/mL

accession no.

NP_004925

UniProt accession no.

storage temp.

−20°C

Gene Information

human ... CDH18(1016)

Application

Coating a plate well (6 well plate) with this recombinant CDH18 protein in neuronal cell specific medium at 1-10 μg/well allows for 1) human neuronal cell / receptor interaction studies or 2) use as a culture matrix protein for human neuronal axon connection studies in vitro.

Use this procedure as a guideline to determine optimal coating conditions for the culture system of choice.
1. Thaw CDH18 and dilute to desired concentration using serum-free medium or PBS. The final solution should be sufficiently dilute so the volume added covers the surface evenly (1-10 μg/well, 6 well plate).
2. Add appropriate amount of diluted material to culture surface.
3. Incubate at room temperature for approximately 1.5 hours.
4. Aspirate remaining material.
5. Rinse plates carefully with water and avoid scratching bottom surface of plates.
6. Plates are ready for use. They may also be stored at 2-8 °C damp or air dried if sterility is maintained.

Sequence

MASMTGGQQMGRGHHHHHHGNLYFQGGEFGWVWNQFFVLEEHMGPDPQYVGKLHSNSDKGDGSVKYILTGEGAGTIFIIDDTTGDIHSTKSLDREQKTHYVLHAQAIDRRTNKPLEPESEFIIKVQDINDNAPKFTDGPYIVTVPEMSDMGTSVLQVTATDADDPTYGNSARVVYSILQGQPYFSVDPKTGVIRTALHNMDREAREHYSVVIQAKDMAGQVGGLSGSTTVNITLTDVNDNPPRFPQKHYQLYVPESAQVGSAVGKIKANDADTGSNADMTYSIINGDGMGIFSISTDKETREGILSLKKPLNYEKKKSYTLNIEGANTHLDFRFSHLGPFKDATMLKIIVGDVDEPPLFSMPSYLMEVYENAKIGTVVGTVLAQDPDSTNSLVRYFINYNVEDDRFFNIDANTGTIRTTKVLDREETPWYNITVTASEIDNPDLLSHVTVGIRVLDVNDNPPELAREYDIIVCENSKPGQVIHTISATDKDDFANGPRFNFFLDERLPVNPNFTLKDNEDNTASILTRRRRFSRTVQDVYYLPIMISDGGIPSLSSSSTLTIRVCACERDGRVRTCHAEAFLSS

Preparation Note

The full-length extracellular domain of the human CDH18 gene (54-608 aa) was constructed with 29 N-terminal T7/HIS-tag and expressed in E. coli as inclusion bodies. The final product was refolded using our unique “temperature shift inclusion body refolding” technology and chromatographically purified.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Oana Viorela Nistor et al.
Molecules (Basel, Switzerland), 25(21) (2020-11-06)
The importance of bioactive compounds such as betalains and anthocyanins was highlighted in the present study by the valorization of red beetroot and sour cherry as an attempt to develop healthy products. The aim of the study was to obtain
Y Shimoyama et al.
The Biochemical journal, 349(Pt 1), 159-167 (2000-06-22)
We identified three novel human type-II classic cadherins, cadherin-7, -9 and -10, by cDNA cloning and sequencing, and confirmed that they interact with catenins and function in cell-cell adhesion as do other classic cadherins. Cell-cell binding activities of the eight
T Shibata et al.
The Journal of biological chemistry, 272(8), 5236-5240 (1997-02-21)
Cadherins, a family of Ca2+-dependent cell-cell adhesion molecules, mediate neural cell-cell interactions and may play important roles in neural development. By searching for molecules that interact with beta-catenin, a cytoplasmic regulator of cadherins, we have identified a new member of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service