Skip to Content
Merck
All Photos(1)

Key Documents

49532

Supelco

Micro particles, magnetic, streptavidin coated

1 μm particle size, std dev <0.1 μm

Synonym(s):

Polystyrene beads, magnetic, streptavidin coated

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.25

grade

analytical standard

form

aqueous solution

composition

iron oxide, ≥20%

concentration

1% (solids)

particle size

1 μm std dev <0.1 μm

application(s)

glass & ceramic
industrial qc
pharmaceutical

format

neat

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

Streptavidin-coated polystyrene-based magnetic microparticles are a size standard, fit for particle size distribution (PSD) analysis of test samples.
The accurate diameter of 1 μm is determined by methods described by the National Institute of Standards and Technology (NIST, USA), such as:
  • Transmission electron microscopy
  • Scanning electron microscopy
  • Optical microscopy
  • COULTER COUNTER particle size analyzer with HDF system.
  • The measuring instruments are calibrated using standards obtained from the Community Bureau of Reference (BCR, Belgium: Standards RM 165, 166 or 167) and from the NIST (SRM 1690, 1691 or 1692). This analytical standard product is made up of superparamagnetic polystyrene particles.

Superparamagnetic polystyrene particles

Application

Easily conjugated with biotin, making them useful for several detection assays,.
Used to calibrate and test particle size analyzers. Easily conjugated with biotin, making it useful for several detection assays.

Features and Benefits

  • suitable for routine instrument calibration checks, testing and corrections
  • available in 5 mL and 10 mL pack sizes as a neat sample

Analysis Note

Binding capacity for FITC-Biotin: >1500 pmol/mg

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Structure and lipid-binding properties of the kindlin-3 pleckstrin homology domain
Ni T, et al.
Journal of Biochemistry, 474(4), 539-556 (2017)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service