Skip to Content
Merck
All Photos(5)

Key Documents

A1645

Sigma-Aldrich

Ammonium phosphate monobasic

ReagentPlus®, ≥98.5%

Synonym(s):

mono-Ammonium phosphate, prim-Ammonium phosphate, Ammonium dihydrogenphosphate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH4H2PO4
CAS Number:
Molecular Weight:
115.03
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.21
grade:
reagent
form:
powder
solubility:
water: soluble(lit.)

grade

reagent

Quality Level

vapor pressure

<0.001 hPa ( 20 °C)

product line

ReagentPlus®

Assay

≥98.5%

form

powder

pH

3.8-4.4 (25 °C, 50 g/L)

mp

190 °C (dec.) (lit.)

solubility

water: soluble(lit.)

SMILES string

N.OP(O)(O)=O

InChI

1S/H3N.H3O4P/c;1-5(2,3)4/h1H3;(H3,1,2,3,4)

InChI key

LFVGISIMTYGQHF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Ammonium phosphate monobasic (APM) is an inorganic salt widely used in fertilizer, baking powder and yeast production. It is also employed as an agent to control pH of a solution and in corrosion inhibition. APM can be prepared by reacting phosphoric acid with anhydrous ammonia maintaining the pH below 5.8. The change in some of the physical characteristics on its addition to a high carbonate containing unfired and fired illitic shale has been studied.

Application


  • Chitoporin from Serratia marcescens: recombinant expression, purification and crystallization.: This study details the application of ammonium phosphate monobasic in the crystallization of chitoporin from Serratia marcescens. The results provide insights into the structural biology of this protein, which is significant for understanding its function and potential applications in biotechnology (Amornloetwattana et al., 2020).



Legal Information

ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Magnetic and luminescent hybrid nanomaterial based on Fe3O4 nanocrystals and GdPO4: Eu3+ nanoneedles.
Runowski M, et al.
Journal of Nanoparticle Research, 14(10), 1-8 (2012)
Development of an inhibitor screening platform via mass spectrometry.
Rathore R, et al.
Journal of Biomolecular Screening, 13(10), 1007-1013 (2008)
Effect of medium components and metabolic inhibitors on β-galactosidase production and secretion by Penicillium notatum 1.
Fiedurek J, et al.
Journal of Basic Microbiology, 36(1), 27-32 (1996)
Development and validation of a method for the determination of EDTA in non-alcoholic drinks by HPLC.
Cagnasso CE, et al.
J. Food Compos. Anal., 20(3), 248-251 (2007)
Burdock GA.
Encyclopedia of Food and Color Additives, 1, 142-142 (1997)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service