Skip to Content
Merck
All Photos(3)

Key Documents

215465

Sigma-Aldrich

Mercury(II) chloride

ACS reagent, ≥99.5%

Synonym(s):

Mercuric chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HgCl2
CAS Number:
Molecular Weight:
271.50
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.21
Assay:
≥99.5%
grade:
ACS reagent
form:
powder
solubility:
acetic acid: soluble(lit.)
acetone: soluble in salt form(lit.)
alcohol: soluble(lit.)
diethyl ether: passes test
ethyl acetate: soluble(lit.)
glycerol: soluble(lit.)
water: soluble(lit.)

grade

ACS reagent

Quality Level

vapor pressure

1.3 mmHg ( 236 °C)

Assay

≥99.5%

form

powder

reaction suitability

reagent type: catalyst
core: mercury

reduction residue

≤0.02%

mp

277 °C (lit.)

solubility

acetic acid: soluble(lit.)
acetone: soluble in salt form(lit.)
alcohol: soluble(lit.)
diethyl ether: passes test
ethyl acetate: soluble(lit.)
glycerol: soluble(lit.)
water: soluble(lit.)

cation traces

Fe: ≤0.002%

SMILES string

Cl[Hg]Cl

InChI

1S/2ClH.Hg/h2*1H;/q;;+2/p-2

InChI key

LWJROJCJINYWOX-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

General description

Mercury (II) chloride is an inorganic salt commonly used as a catalyst in organic synthesis.

Application

Mercury (II) chloride can be used as a catalyst for the:
  • Addition reaction between aromatic amines to terminal acetylenes to give imines, enamines, and 1,2,3,4-tetrahydroquinoline derivatives.
  • Cyclization of amidinothioureas with phenyl hydrazine to produce 3-amino-1,2,4-triazole derivatives.
  • Cyclization of O-propargyl glycolaldehyde dithioacetals to afford 3-pyranones along with 2,5-dihydrofuran-3-carboxaldehydes through their dithioketals and dithioacetals reaction.
  • Reductive dimerization and cyclization of α,β-unsaturated ketones produce functionalized cyclopentanols using N,N-dimethylformamide as solvent.

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Muta. 2 - Repr. 2 - Skin Corr. 1B - STOT RE 1

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ashley E Peppriell et al.
Toxicology, 443, 152561-152561 (2020-08-18)
Methylmercury (MeHg) is a ubiquitous environmental contaminant and developmental toxicant known to cause a variety of persistent motor and cognitive deficits. While previous research has focused predominantly on neurotoxic MeHg effects, emerging evidence points to a myotoxic role whereby MeHg
Mercury(II) Chloride.
Koc ovsky P.
e-EROS Encyclopedia of Reagents for Organic Synthesis. (2001)
Eagleson M.
Concise Encyclopedia Chemistry, 498-498 (1994)
Vasco Branco et al.
Free radical biology & medicine, 52(4), 781-793 (2011-12-27)
Mercury compounds exert toxic effects via interaction with many vital enzymes involved in antioxidant regulation, such as selenoenzymes thioredoxin reductase (TrxR) and glutathione peroxidase (GPx). Selenium supplementation can reactivate the mercury-inhibited TrxR and recover the cell viability in vitro. To
Igor Shuryak et al.
Scientific reports, 9(1), 11361-11361 (2019-08-08)
Exposure to chronic ionizing radiation (CIR) from nuclear power plant accidents, acts of terrorism, and space exploration poses serious threats to humans. Fungi are a group of highly radiation-resistant eukaryotes, and an understanding of fungal CIR resistance mechanisms holds the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service