Skip to Content
Merck
All Photos(1)

Key Documents

Y0000845

Atropine for peak identification

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Atropine, Hyoscyamine, Tropine tropate, endo-(±)-α-(Hydroxymethyl)benzeneacetic acid 8-methyl-8-azabicyclo[3.2.1]oct-3-yl ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H23NO3
CAS Number:
Molecular Weight:
289.37
Beilstein:
91260
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

atropine

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

CN1[C@H]2CC[C@@H]1C[C@@H](C2)OC(=O)C(CO)c3ccccc3

InChI

1S/C17H23NO3/c1-18-13-7-8-14(18)10-15(9-13)21-17(20)16(11-19)12-5-3-2-4-6-12/h2-6,13-16,19H,7-11H2,1H3/t13-,14+,15+,16?

InChI key

RKUNBYITZUJHSG-SPUOUPEWSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Atropine for peak identification EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Biochem/physiol Actions

Competitive nonselective antagonist at central and peripheral muscarinic acetylcholine receptors.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 2 Oral

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Joanna Kur et al.
The Journal of physiology, 592(3), 491-504 (2013-11-28)
Purinergic control of vascular tone in the CNS has been largely unexplored. This study examines the contribution of endogenous extracellular ATP, acting on vascular smooth muscle cells, in controlling vascular tone in the in vivo rat retina. Retinal vessels were
Michela Faggioni et al.
Circulation research, 112(4), 689-697 (2013-01-09)
Catecholaminergic polymorphic ventricular tachycardia (CPVT) is caused by mutations in cardiac ryanodine receptor (RyR2) or calsequestrin (Casq2) genes. Sinoatrial node dysfunction associated with CPVT may increase the risk for ventricular arrhythmia (VA). To test the hypothesis that CPVT is suppressed
Incidence, pathophysiology, and treatment of complications during dobutamine-atropine stress echocardiography.
Marcel L Geleijnse et al.
Circulation, 121(15), 1756-1767 (2010-04-21)
Peter Jones et al.
Archives of disease in childhood, 97(2), 139-144 (2011-05-31)
Bradycardia occurs during the intubation of some critically ill children as a result of vagal stimulation due to hypoxia and/or laryngeal stimulation; such 'stable' bradycardia is accompanied by selective vasoconstriction. Some induction drugs also induce bradycardia which may be accompanied
D Poldermans et al.
European heart journal, 18 Suppl D, D86-D96 (1997-06-01)
Cardiac events in the peri-operative phase and late after non-cardiac vascular surgery are a major cause of morbidity and mortality. Numerous tests and diagnostic strategies--usually consisting of a combination of analysis of clinical risk factors and additional non-exercise dependent stress

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service