Recommended Products
grade
certified reference material
manufacturer/tradename
(METAS)
application(s)
food and beverages
format
matrix material
General description
More detailed information about the development of this product can be found in this paper published in july 2023 in Analytical and Bioanalytical Chemistry
Other Notes
Trace Elements: Arsenic (As), Cadmium (Cd), Lead (Pb), Mercury (Hg)
Organic Pollutants: Benz[a]anthracene, Benzo[a]pyrene, Benzo[b]fluoranthene, Chrysene
Matrix Group: Dairy products and eggs
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service