Skip to Content
Merck
All Photos(1)

Key Documents

S0050000

Saccharin sodium

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Saccharin sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H4NNaO3S
CAS Number:
Molecular Weight:
205.17
UNSPSC Code:
41116107
NACRES:
NA.24
Pricing and availability is not currently available.

grade

pharmaceutical primary standard

API family

saccharin

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

[Na+].[S]1(=NC(=O)c2c1cccc2)(=O)[O-]

InChI

1S/C7H5NO3S.Na/c9-7-5-3-1-2-4-6(5)12(10,11)8-7;/h1-4H,(H,8,9,10,11);/q;+1/p-1

InChI key

OAZGZMKGRPRHJX-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Saccharin sodium EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

428.0 °F - closed cup

Flash Point(C)

220 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Heather E King et al.
Pharmacology, biochemistry, and behavior, 126, 163-169 (2014-10-07)
The inbred Fischer (F344) and Lewis (LEW) rats, while originally developed as animal models for cancer and tissue transplantation research, have since been used to study genetic differences in a variety of physiological and behavioral endpoints. In this context, LEW
Yangjie Wei et al.
AAPS PharmSciTech, 16(2), 455-465 (2014-11-02)
Several attempts have been made to mask the bitter taste of oral formulations, but none have been made for injectable formulations. This study aims to mask the bitter taste of dental lidocaine HCl (LID) injection using hydroxypropyl-β-cyclodextrin (HP-β-CD) and sodium
Anthony Sclafani et al.
American journal of physiology. Regulatory, integrative and comparative physiology, 307(12), R1448-R1457 (2014-10-17)
Recent studies indicate that, unlike glucose, fructose has little or no post-oral preference conditioning actions in C57BL/6J (B6) mice. The present study determined whether this is also the case for FVB mice, which overconsume fructose relative to B6 mice. In

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service