Skip to Content
Merck
All Photos(2)

Documents

RDD042

Sigma-Aldrich

Sodium 1-heptanesulfonate

Redi-Dri, free-flowing

Synonym(s):

1-Heptanesulfonic acid sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C7H15O3SNa
CAS Number:
Molecular Weight:
202.25
MDL number:
UNSPSC Code:
12161900
NACRES:
NA.21

product line

Redi-Dri

Quality Level

quality

free-flowing

SMILES string

[Na+].CCCCCCCS([O-])(=O)=O

InChI

1S/C7H16O3S.Na/c1-2-3-4-5-6-7-11(8,9)10;/h2-7H2,1H3,(H,8,9,10);/q;+1/p-1

InChI key

REFMEZARFCPESH-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Sodium 1-heptanesulfonate belongs to the sodium alkylsulfonate group of surfactants. It has a -SO₃ head group with sodium and an alkyl chain attached to the head group. is Alkylsofonates are often used as a ion pairing reagents for ICP the analysis of cations.

Application

Redi-Dri is an innovative packaging product line that helps eliminate clumping material by adding extra packaging process steps without any chemical additives, anti-clumping agents, or hydrophobic compounds. Redi-Dri offers a more convenient and safer way to handle the products.
Sodium 1-heptanesulfonate has been used:
  • in mobile phase buffer for preequilibration of High Performance Liquid Chromatography (HPLC) column.
  • Ion-pairing reagent for HPLC to analyze peptides and proteins
  • ion pair reagent for analysis of epinephrine and norepinephrine in ion-pairing chromatography (IPC).

Ion-pairing reagent for HPLC, including analyses of peptides and proteins.

Features and Benefits

  • Redi-Dri packaging controls moisture for hygroscopic chemicals that commonly clump
  • Anhydrous powder remains free flowing making it easier to use
  • Safer to handle and weigh out for routine use in lab
  • Eliminates clumps, maintaining product moisture, high quality attributes

Other Notes

For additional information on our range of Biochemicals, please complete this form.

Legal Information

Redi-Dri is a trademark of Sigma-Aldrich Co. LLC

comparable product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 2


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Oshizaka T, Kikuchi K, Kadhum WR, et al.
International Journal of Pharmaceutical and Chemical Sciences, 475(1-2), 292-297 (2014)
EURASIP journal on bioinformatics & systems biology, 88(2), 510-517 (2014)
Analysis of catecholamines in urine by unique LC/MS suitable ion-pairing chromatography
Bergmann ML, et al.
Journal of Chromatography. B, Analytical Technologies in the Biomedical and Life Sciences, 1057(2), 118-123 (2017)
Validated HPLC-Fl method for the analysis of S-adenosylmethionine and S-adenosylhomocysteine biomarkers in human blood
Albu C, et al.
Journal of Fluorescence, 23(3), 381-386 (2013)
Determination of S-adenosylmethionine and S-adenosylhomocysteine from human blood samples by HPLC-FL
Birsan C, et al.
Analytical Letters, 41(10), 1720-1731 (2008)

Articles

Redi-Dri™ prevents hygroscopic powders, such as inorganic salts, from absorbing moisture and forming clumps, leaving the salts free-flowing every time.

Redi-Dri™ prevents hygroscopic powders, such as inorganic salts, from absorbing moisture and forming clumps, leaving the salts free-flowing every time.

Redi-Dri™ prevents hygroscopic powders, such as inorganic salts, from absorbing moisture and forming clumps, leaving the salts free-flowing every time.

Redi-Dri™ prevents hygroscopic powders, such as inorganic salts, from absorbing moisture and forming clumps, leaving the salts free-flowing every time.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service