Skip to Content
Merck
All Photos(1)

Key Documents

PHR3825

Supelco

Ciprofloxacin Impurity C

certified reference material, pharmaceutical secondary standard

Synonym(s):

7-[(2-Aminoethyl)amino]-1-cyclopropyl-6-fluoro-1,4-dihydro-4-oxo-3-quinolinecarboxylic acid hydrochloride, Ciprofloxacin Ethylenediamine Analog, Desethylene Ciprofloxacin, Desethylene Ciprofloxacin HCl

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H16FN3O3 · HCl
CAS Number:
Molecular Weight:
341.77
UNSPSC Code:
41116107

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

traceable to USP 1134324

API family

ciprofloxacin

packaging

pkg of 30MG

application(s)

pharmaceutical

storage temp.

2-30°C

InChI

1S/C15H16FN3O3.ClH/c16-11-5-9-13(6-12(11)18-4-3-17)19(8-1-2-8)7-10(14(9)20)15(21)22;/h5-8,18H,1-4,17H2,(H,21,22);1H

InChI key

ODOFTUIHVVTJRI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Pharmaceutical secondary standards for application in quality control provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards

Analysis Note

These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service