Skip to Content
Merck
All Photos(1)

Key Documents

PHR1372

Supelco

Methanol

Pharmaceutical Secondary Standard; Certified Reference Material

Synonym(s):

Methanol, Methyl alcohol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3OH
CAS Number:
Molecular Weight:
32.04
Beilstein:
1098229
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

traceable to USP 1424109

vapor density

1.11 (vs air)

vapor pressure

410 mmHg ( 50 °C)
97.68 mmHg ( 20 °C)

CofA

current certificate can be downloaded

autoignition temp.

725 °F

expl. lim.

36 %

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.329 (lit.)

bp

64.7 °C (lit.)

mp

−98 °C (lit.)

density

0.791 g/mL at 25 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-30°C

SMILES string

CO

InChI

1S/CH4O/c1-2/h2H,1H3

InChI key

OKKJLVBELUTLKV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Pharmaceutical secondary standards for application in quality control provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards. Methanol is a widely used polar organic solvent in chemistry, biochemistry, and molecular biology. It is a common solvent in HPLC and mass spectrometry (MS) of small molecules and of biomolecules such as oligonucleotides and proteins. Methanol is also utilized in high performance capillary electrophoresis. Industrial applications of methanol include the manufacture of formaldehyde and methyl esters of organic and inorganic acids. Methanol is used as a solvent and solvent adjuvant for polymers, and as a softening agent for pyroxylin plastics.

Application

These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.

Footnote

To see an example of a Certificate of Analysis for this material enter LRAB7353 in the slot below. This is an example certificate only and may not be the lot that you receive.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hany K M Dweck et al.
Current biology : CB, 25(4), 455-466 (2015-01-27)
Dietary antioxidants play an important role in preventing oxidative stress. Whether animals in search of food or brood sites are able to judge the antioxidant content, and if so actively seek out resources with enriched antioxidant content, remains unclear. We
Lakshmi Gopinathan et al.
Cancer research, 74(14), 3870-3879 (2014-05-08)
Cell-cycle inhibition has yet to offer a generally effective approach to cancer treatment, but a full evaluation of different combinations of cell-cycle inhibitors has not been evaluated. Cyclin A2, a core component of the cell cycle, is often aberrantly expressed
Timo Heidt et al.
Circulation research, 115(2), 284-295 (2014-05-03)
Macrophages populate the steady-state myocardium. Previously, all macrophages were thought to arise from monocytes; however, it emerged that, in several organs, tissue-resident macrophages may self-maintain through local proliferation. Our aim was to study the contribution of monocytes to cardiac-resident macrophages
Nicolas Vodovar et al.
European heart journal, 35(48), 3434-3441 (2014-08-27)
Increases in plasma B-type natriuretic peptide (BNP) concentrations in those with acutely decompensated heart failure (ADHF) has been mainly attributed to an increase in NPPB gene transcription. Recently, proBNP glycosylation has emerged as a potential regulatory mechanism in the production
B Alkhatib et al.
European cells & materials, 28, 98-110 (2014-09-13)
Excessive mechanical loading or acute trauma to intervertebral discs (IVDs) is thought to contribute to degeneration and pain. However, the exact mechanisms by which mechanical injury initiates and promotes degeneration remain unclear. This study investigates biochemical changes and extracellular matrix

Articles

Analysis of carbonate solvents in lithium-ion batteries like ethylene carbonate, dimethyl carbonate, diethyl carbonate, and more using SPB®-624 column.

Analysis of carbonate solvents in lithium-ion batteries like ethylene carbonate, dimethyl carbonate, diethyl carbonate, and more using SPB®-624 column.

Analysis of carbonate solvents in lithium-ion batteries like ethylene carbonate, dimethyl carbonate, diethyl carbonate, and more using SPB®-624 column.

Analysis of carbonate solvents in lithium-ion batteries like ethylene carbonate, dimethyl carbonate, diethyl carbonate, and more using SPB®-624 column.

Chromatograms

suitable for GC

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service