Skip to Content
Merck
All Photos(1)

Key Documents

PHR1337

Supelco

Thiamine Nitrate

Pharmaceutical Secondary Standard; Certified Reference Material

Synonym(s):

Thiamine nitrate, Vitamin B1 nitrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H17N5O4S
CAS Number:
Molecular Weight:
327.36
Beilstein:
3857790
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

traceable to BP 414
traceable to Ph. Eur. Y0000060

API family

thiamine

CofA

current certificate can be downloaded

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

[O-][N+]([O-])=O.CC1=NC=C(C[N+]2=CSC(CCO)=C2C)C(N)=N1

InChI

1S/C12H17N4OS.NO3/c1-8-11(3-4-17)18-7-16(8)6-10-5-14-9(2)15-12(10)13;2-1(3)4/h5,7,17H,3-4,6H2,1-2H3,(H2,13,14,15);/q+1;-1

InChI key

UIERGBJEBXXIGO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Certified pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards. Thiamine Nitrate is a mononitrate salt of thiamine. It is synthesized from thiamine hydrochloride following a sequence of steps.[1] Thiamine Nitrate is used as a dietary supplement in crackers, eggs substitute and enriched flour.[2]

Application

These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.

Analysis Note

These secondary standards offer multi-traceability to the USP, EP (PhEur) and BP primary standards, where they are available.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Footnote

To see an example of a Certificate of Analysis for this material enter LRAC3016 in the slot below. This is an example certificate only and may not be the lot that you receive.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Food Additives Handbook (1989)
Encyclopedia of Food and Color Additives, 3 (1997)
Encyclopedia of Food and Color Additives, 3 (1997)
Gloria Uccello-Barretta et al.
Journal of pharmaceutical and biomedical analysis, 107, 7-10 (2015-01-07)
In total parenteral nutrition (TPN), especially in the case of preterm infants, simultaneous administration of vitamins and trace elements is still a problematic issue: guidelines put in evidence the lack of specific documentation. In this work NMR spectroscopy was applied

Articles

Learn to separate and quantify B vitamins and preservatives in capsules with HPLC-UV and Purospher® STAR RP-8 column. Validated for precision.

Learn to separate and quantify B vitamins and preservatives in capsules with HPLC-UV and Purospher® STAR RP-8 column. Validated for precision.

Learn to separate and quantify B vitamins and preservatives in capsules with HPLC-UV and Purospher® STAR RP-8 column. Validated for precision.

Learn to separate and quantify B vitamins and preservatives in capsules with HPLC-UV and Purospher® STAR RP-8 column. Validated for precision.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service