Skip to Content
Merck
All Photos(1)

Key Documents

PHR1056

Supelco

Trimethoprim

Pharmaceutical Secondary Standard; Certified Reference Material

Synonym(s):

2,4-Diamino-5-(3,4,5-trimethoxybenzyl)pyrimidine, NSC 106568

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H18N4O3
CAS Number:
Molecular Weight:
290.32
Beilstein:
625127
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

traceable to BP 344
traceable to Ph. Eur. T220000
traceable to USP 1692505

API family

trimethoprim

CofA

current certificate can be downloaded

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-30°C

SMILES string

COc1cc(Cc2cnc(N)nc2N)cc(OC)c1OC

InChI

1S/C14H18N4O3/c1-19-10-5-8(6-11(20-2)12(10)21-3)4-9-7-17-14(16)18-13(9)15/h5-7H,4H2,1-3H3,(H4,15,16,17,18)

InChI key

IEDVJHCEMCRBQM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Certified pharmaceutical secondary standards for application in quality control provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to in-house working standards. Trimethoprim is a synthetic antibacterial agent active in vitro against most aerobic gram-negative and gram-positive bacteria. Its mode of action involves inhibition of dihydrofolate reductase, an enzyme involved in catalyzing the reduction of dihydrofolate to tetrahydrofolate in both microbial and eukaryotic cells.

Application

Primarily used as an antibacterial agent. Dihydrofolate reductase inhibitor with selectivity for the prokaryote enzyme.
These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.
Trimethoprim may be used as a pharmaceutical reference standard for the determination of the analyte in pharmaceutical formulations by chromatography and visible and UV spectrophotometry.

Analysis Note

These secondary standards offer multi-traceability to the USP, EP (PhEur) and BP primary standards, where they are available.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Footnote

To see an example of a Certificate of Analysis for this material enter LRAA7172 in the slot below. This is an example certificate only and may not be the lot that you receive.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Repr. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Comparison of trimethoprim molecularly imprinted polymers in bulk and in sphere as the sorbent for solid-phase extraction and extraction of trimethoprim from human urine and pharmaceutical tablet and their determination by high-performance liquid chromatography
Hu SG, et al.
Analytica Chimica Acta, 537(1-2), 215-222 (2005)
Trimethoprim resistance.
Huovinen P
Antimicrobial Agents and Chemotherapy, 31(10), 1451-1456 (1987)
Determination of sulfamethoxazole and trimethoprim in pharmaceuticals by visible and UV spectrophotometry
Shamsa F and Amani L
Iranian Journal of Pharmaceutical Research : IJPR, 537(1-2), 31-36 (2010)
Simultaneous LC determination of trimethoprim and sulphamethoxazole in pharmaceutical formulations
Akay C and Ozkan SA
Journal of Pharmaceutical and Biomedical Analysis, 30(4), 1207-1213 (2002)
E Van Duijkeren et al.
Journal of veterinary pharmacology and therapeutics, 17(1), 64-73 (1994-02-01)
The indications for use, side-effects, and pharmacokinetic parameters of trimethoprim, sulfonamides and their combinations in the horse are reviewed. Trimethoprim/sulfonamide (TMPS) combinations are used for the treatment of various diseases caused by gram-positive and gram-negative bacteria, including infections of the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service