Skip to Content
Merck
All Photos(3)

Key Documents

P0597

Sigma-Aldrich

Pilocarpine nitrate

meets USP testing specifications

Synonym(s):

Pilocarpine nitrate salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H16N2O2 · HNO3
CAS Number:
Molecular Weight:
271.27
Beilstein:
4171406
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.21

Agency

USP/NF
meets USP testing specifications

Quality Level

form

crystals

application(s)

pharmaceutical (small molecule)

SMILES string

O[N+]([O-])=O.CC[C@H]1[C@H](COC1=O)Cc2cncn2C

InChI

1S/C11H16N2O2.HNO3/c1-3-10-8(6-15-11(10)14)4-9-5-12-7-13(9)2;2-1(3)4/h5,7-8,10H,3-4,6H2,1-2H3;(H,2,3,4)/t8-,10-;/m0./s1

InChI key

PRZXEPJJHQYOGF-GNAZCLTHSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Nonselective muscarinic acetylcholine receptor agonist.
Nonselective muscarinic acetylcholine receptor agonist; used to produce an experimental model of epilepsy.

Pictograms

Flame over circleSkull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 4 Oral - Ox. Sol. 2

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Olav B Smeland et al.
Journal of cerebral blood flow and metabolism : official journal of the International Society of Cerebral Blood Flow and Metabolism, 33(7), 1090-1097 (2013-04-25)
Although certain metabolic characteristics such as interictal glucose hypometabolism are well established for temporal lobe epilepsy (TLE), its pathogenesis still remains unclear. Here, we performed a comprehensive study of brain metabolism in a mouse model of TLE, induced by pilocarpine-status
Linda Holtman et al.
Epilepsia, 54(4), 589-595 (2013-02-13)
Brain inflammation occurs during epileptogenesis and may contribute to the development and progression of temporal lobe epilepsy. Recently, several studies have indicated that seizures may also increase specific blood plasma cytokine levels in animal models as well as in human
Shuo-Bin Jou et al.
Seizure, 22(3), 221-229 (2013-01-15)
Bilateral electrical stimulation of anterior nuclei of thalamus (ANT) has shown promising effects on epileptic seizures. However, bilateral implantation increases the risk of surgical complications and side effects. This study was undertaken to access the effectiveness of a stimulation paradigm
Kathleen Heng et al.
Epilepsia, 54(9), 1535-1541 (2013-07-16)
The role of granule cell axon (mossy fiber) sprouting in temporal lobe epileptogenesis is unclear and controversial. Rapamycin suppresses mossy fiber sprouting, but its reported effects on seizure frequency are mixed. The present study used high-dose rapamycin to more completely
Manuela Mazzuferi et al.
Annals of neurology, 74(4), 560-568 (2013-05-21)
Epigenetic mechanisms involved in transcriptional regulation of multiple molecular pathways are potentially attractive therapeutic interventions for epilepsy, because single target therapies are unlikely to provide both anticonvulsant and disease-modifying effects. A selection of epilepsy-related gene expression data sets were retrieved

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service