NIST660C
Line position and line shape standard for powder diffraction
NIST® SRM® 660c, Lanthanum hexaboride powder
Synonym(s):
Lanthanum hexaboride, Lanthanum boride
About This Item
Recommended Products
grade
certified reference material
Quality Level
form
powder
packaging
pkg of 6 g
manufacturer/tradename
NIST®
technique(s)
diffraction/scattering: suitable
mp
2210 °C
density
2.61 g/cm3 at 25 °C (lit.)
SMILES string
[La++].B12B3[B-]14B5B4[B-]235
InChI
1S/B6.La/c1-2-5(1)3-4(5)6(1,2)3;/q-2;+2
InChI key
VJXOKIULCJPJLW-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
For more information, please refer to the COA and SDS.
SRM 660C_CERT
SRM 660C_SDS
Application
Features and Benefits
- A unit of SRM 660c consists of approximately 6 g of lanthanum hexaboride, LaB6, powder bottled under argon gas
- Information on product description, disposal, and transportation is available in the accompanying MSDS
- The enclosed NIST certificate contains details on the expiry date, warning to users, instructions for right storage and use, preparation, and analysis of the RM
- Certified values and uncertainties calculated according to the method described in the ISO/JCGM Guide
- Metrological traceability is to the SI unit for length (expressed as nanometers)
Other Notes
Lanthanum Hexaboride Powder
See certificate for values and more details at nist.gov/SRM.
Legal Information
related product
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service