Skip to Content
Merck
All Photos(1)

Documents

NIST1845A

Whole egg powder

NIST® SRM® 1845a

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

Quality Level

packaging

pkg of 5 pouches x 10 g ea

manufacturer/tradename

NIST®

application(s)

food and beverages

format

matrix material

General description

A unit of SRM 1845A consists of five heat-sealed aluminized pouches, each pouch contains ~10 g of material.

SRM 1845A_cert

SRM 1845A_SDS

Application

NIST1845A Whole egg powder standard reference material (SRM) is intended primarily for validation of methods for determining proximate, fatty acids, cholesterol, vitamins, elements, and amino acids in whole egg powder.

Features and Benefits

  • Available with a certificate documenting NIST-certified mass fraction values cholesterol, elements, and vitamins. 
  • Reference values are expressed as mass fractions for other additional elements. 
  • Certified values are metrologically traceable to the SI measurement unit for chemical mass fraction, expressed as milligrams per gram.  
  • The NIST certificate is provided with storage and use instructions.

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service