Skip to Content
Merck
All Photos(1)

Documents

I0020000

Ibuprofen

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

α-Methyl-4-(isobutyl)phenylacetic acid, (±)-2-(4-Isobutylphenyl)propanoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H18O2
CAS Number:
Molecular Weight:
206.28
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

ibuprofen

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

CC(C)Cc1ccc(cc1)C(C)C(O)=O

InChI

1S/C13H18O2/c1-9(2)8-11-4-6-12(7-5-11)10(3)13(14)15/h4-7,9-10H,8H2,1-3H3,(H,14,15)

InChI key

HEFNNWSXXWATRW-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Ibuprofen EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Biochem/physiol Actions

Cyclooxygenase (COX) inhibitor that has greater activity against COX-1 than against COX-2.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

related product

Product No.
Description
Pricing

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J B Whitlam et al.
Clinical pharmacology and therapeutics, 29(4), 487-492 (1981-04-01)
Free and total ibuprofen levels in serum and synovial fluid (SF) were determined in one male and 14 female arthritic patients (mean age 56 yr, range 19 to 77) after 400 mg three times daily for 2 days. Free drug
Sheena Derry et al.
The Cochrane database of systematic reviews, 10(10), CD007550-CD007550 (2013-10-24)
This review is an update of a previously published review in The Cochrane Database of Systematic Reviews Issue 3, 2009 on single dose oral dexibuprofen (S(+)-ibuprofen) for acute postoperative pain in adults.Dexibuprofen is a non-steroidal anti-inflammatory drug (NSAID) licensed for
Massimo Imazio et al.
The New England journal of medicine, 369(16), 1522-1528 (2013-09-03)
Colchicine is effective for the treatment of recurrent pericarditis. However, conclusive data are lacking regarding the use of colchicine during a first attack of acute pericarditis and in the prevention of recurrent symptoms. In a multicenter, double-blind trial, eligible adults
Bart Van Overmeire et al.
Lancet (London, England), 364(9449), 1945-1949 (2004-11-30)
Ibuprofen is used for treatment and prevention of patent ductus arteriosus in low-birthweight infants. Its effects on regional circulations differ from those of indometacin. Because prophylactic indometacin reduces the frequency of severe intraventricular haemorrhage and patent ductus arteriosus, we aimed
R O Day et al.
Clinical pharmacology and therapeutics, 43(5), 480-487 (1988-05-01)
The simultaneous disposition of the enantiomers of ibuprofen in synovial fluid and plasma was studied in eight patients with arthritis. Concentrations of the active S-enantiomer in synovial fluid exceeded those of the R-enantiomer at all times in all patients with

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service