Skip to Content
Merck
All Photos(1)

Documents

BP248

Methylprednisolone

British Pharmacopoeia (BP) Reference Standard

Synonym(s):

6α-Methylprednisolone, 11β,17α,21-Trihydroxy-6α-methyl-1,4-pregnadiene-3,20-dione, 6α-Methyl-11β,17α,21-trihydroxy-1,4-pregnadiene-3,20-dione, Medrol, Medrone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H30O5
CAS Number:
Molecular Weight:
374.47
Beilstein:
2340300
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

methylprednisolone

form

powder

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

BP

application(s)

pharmaceutical
pharmaceutical small molecule

format

neat

storage temp.

2-8°C

SMILES string

[H][C@@]12C[C@H](C)C3=CC(=O)C=C[C@]3(C)[C@@]1([H])[C@@H](O)C[C@@]4(C)[C@@]2([H])CC[C@]4(O)C(=O)CO

InChI

1S/C22H30O5/c1-12-8-14-15-5-7-22(27,18(26)11-23)21(15,3)10-17(25)19(14)20(2)6-4-13(24)9-16(12)20/h4,6,9,12,14-15,17,19,23,25,27H,5,7-8,10-11H2,1-3H3/t12-,14-,15-,17-,19+,20-,21-,22-/m0/s1

InChI key

VHRSUDSXCMQTMA-PJHHCJLFSA-N

Gene Information

human ... NR3C1(2908)

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Methylprednisolone BP Reference standard, intended for use in laboratory tests only as specifically prescribed in the British Pharmacopoeia.

Packaging

Unit quantity: 100 mg. Subject to change. The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity please visit British Pharmacopoeia

Other Notes

Sales restrictions may apply.

related product

Product No.
Description
Pricing

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Repr. 1B - STOT RE 2

Target Organs

Adrenal gland,Immune system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Jingze Zang et al.
Current biology : CB, 31(6), 1251-1260 (2021-02-17)
In plants, the cortical endoplasmic reticulum (ER) network is connected to the plasma membrane (PM) through the ER-PM contact sites (EPCSs), whose structures are maintained by EPCS resident proteins and the cytoskeleton.1-7 Strong co-alignment between EPCSs and the cytoskeleton is
Vanessa Silva Silveira et al.
Scientific reports, 10(1), 15193-15193 (2020-09-18)
SHOC2 scaffold protein has been mainly related to oncogenic ERK signaling through the RAS-SHOC2-PP1 phosphatase complex. In leukemic cells however, SHOC2 upregulation has been previously related to an increased 5-year event-free survival of pediatric pre-B acute lymphoid leukemia, suggesting that

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service