BCR339
6H-Benzo[cd]pyren-6-one
BCR®, certified reference material
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C19H10O
CAS Number:
Molecular Weight:
254.28
Beilstein:
2052940
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24
grade
certified reference material
Agency
BCR®
manufacturer/tradename
JRC
technique(s)
HPLC: suitable
gas chromatography (GC): suitable
format
neat
storage temp.
2-8°C
SMILES string
O=C1c2cccc3ccc4ccc5cccc1c5c4c23
InChI
1S/C19H10O/c20-19-14-5-1-3-11-7-9-13-10-8-12-4-2-6-15(19)17(12)18(13)16(11)14/h1-10H
InChI key
CLIKSBRDCNSYNO-UHFFFAOYSA-N
Analysis Note
For more information please see:
BCR339
BCR339
Legal Information
BCR is a registered trademark of European Commission
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service