Skip to Content
Merck
All Photos(1)

Key Documents

A0152000

Acetylcysteine impurity C

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

N,N′-Diacetyl-L-cystine, NSC 203780

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H16N2O6S2
CAS Number:
Molecular Weight:
324.37
EC Number:
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24
Pricing and availability is not currently available.

grade

pharmaceutical primary standard

API family

acetylcysteine

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

S(SCC(NC(=O)C)C(=O)O)CC(NC(=O)C)C(=O)O

InChI

1S/C10H16N2O6S2/c1-5(13)11-7(9(15)16)3-19-20-4-8(10(17)18)12-6(2)14/h7-8H,3-4H2,1-2H3,(H,11,13)(H,12,14)(H,15,16)(H,17,18)

InChI key

YTPQSLLEROSACP-UHFFFAOYSA-N

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets, has been developed and issued under the Authority of the issuing Pharmacopoeia.

For further information and support please go to the website of the issuing Pharmacopoeia.

Application

This European Pharmacopoeia reference standard is intended for use only as specifically prescribed in the European Pharmacopoeia. Their suitability for any other use is not guaranteed and is the sole responsibility of the user. This standard is not intended for human or animal use.
Established for the preparation of reference solution (c) in testing for related substances in acetylcysteine using liquid chromatography (General text 2.2.29), according to the monograph 0967 of European Pharmacopeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Acetylcysteine
European Pharmacopoeia Commission and European Directorate for the Quality of Medicines & Healthcare
European pharmacopoeia, 4900-4901 (2021)
Knut Pettersson et al.
Cardiovascular drug reviews, 21(2), 119-132 (2003-07-09)
Inflammatory processes in the arterial wall are important in atherogenesis. The present review discusses the development of DiNAC as a potential new treatment modality for atherosclerosis related diseases. DiNAC, N,N'-diacetyl-L-cystine, is the disulphide dimer of N-acetyl cysteine, NAC. It was
Sheila A Doggrell
Expert opinion on investigational drugs, 11(5), 717-720 (2002-05-09)
Not all antioxidants reduce atherosclerosis. This may be because atherosclerosis has an autoimmune, inflammatory pathogenesis. As probucol is both an antioxidant and an immunomodulatory drug, it may be the immunomodulatory effect that underlies its ability to reduce atherosclerosis. N,N-Diacetyl-L-cystine is
S Boräng et al.
Gene, 271(2), 183-192 (2001-06-22)
In this study, we have applied and evaluated a modified cDNA representational difference analysis (RDA) protocol based on magnetic bead technology to study the molecular effects of a candidate drug (N,N'-diacetyl-L-cystine, DiNAC) in a model for atherosclerosis. Alterations in a
S Böhler et al.
Infusionstherapie (Basel, Switzerland), 15(2), 89-92 (1988-04-01)
In this study the question of whether N-N-diacetylcystine (DAC), which is more stable than N-acetylcysteine (AcCYS), may provide a useful cysteine source for parenteral nutrition was investigated. In in vitro studies the release of cysteine from DAC was measured. The

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service