Skip to Content
Merck
All Photos(1)

Key Documents

89067

Supelco

Corosolic acid

analytical standard

Synonym(s):

(2α,3β)-2,3-Dihydroxyurs-12-en-28-oic acid, 2α-Hydroxyursolic acid, Colosolic acid, Corsolic acid, Glucosol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C30H48O4
CAS Number:
Molecular Weight:
472.70
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥85.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

food and beverages

format

neat

SMILES string

O[C@H]1[C@H](O)C(C)(C)[C@@](CC[C@]2(C)[C@]3([H])CC=C4[C@@]2(C)CC[C@]5(C(O)=O)[C@@]4([H])[C@@H](C)[C@H](C)CC5)([H])[C@]3(C)C1

InChI

1S/C30H48O4/c1-17-10-13-30(25(33)34)15-14-28(6)19(23(30)18(17)2)8-9-22-27(5)16-20(31)24(32)26(3,4)21(27)11-12-29(22,28)7/h8,17-18,20-24,31-32H,9-16H2,1-7H3,(H,33,34)/t17-,18+,20-,21+,22-,23+,24+,27+,28-,29-,30+/m1/s1

InChI key

HFGSQOYIOKBQOW-ZSDYHTTISA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

Corosolic acid may be used as a reference standard in the determination of corosolic acid in Eriobotrya japonica leaves using reversed-phase high performance liquid chromatography (RP-HPLC).
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Biochem/physiol Actions

Triterpene phytochemical found in medicinal herbal extracts. Possesses antiatherosclerotic, antihyperlipidemic, antioxidant, antiinflammatory, antifungal, antiviral and antineoplastic activities.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Determination of corosolic acid in Eriobotrya japonica leaves by reversed-phase high performance liquid chromatography.
CH, et al.
Se Pu / Chinese Journal of Chromatography, 24(5), 492-494 (2006)
Ke Wang et al.
Oncology reports, 39(6), 2777-2786 (2018-04-06)
Retinoblastoma (Rb) is the most frequent primary intraocular tumor usually diagnosed in infants and children, and current therapy for such disease is still limited. Corosolic acid (CA), an ursane-type pentacyclic triterpene, has been assessed as a promising anticancer agent with little impact
Sidney J Stohs et al.
Phytotherapy research : PTR, 26(3), 317-324 (2011-11-19)
Banaba (Lagerstroemia speciosa L.) extracts have been used for many years in folk medicine to treat diabetes, with the first published research study being reported in 1940. This review summarizes the current literature regarding banaba and its constituents. The hypoglycemic
Baohong Li et al.
Pakistan journal of pharmaceutical sciences, 27(3), 511-516 (2014-05-09)
A sensitive and robust liquid chromatography-tandem mass spectrometry (LC-MS/MS) method was developed and validated for the determination of corosolic acid (CRA) in rat plasma. The plasma samples were prepared by a simple protein precipitation treatment. Chromatographic separation was performed on
Ganapathy Sivakumar et al.
Biotechnology journal, 4(12), 1704-1711 (2009-12-01)
Diabetes is one of the nation's most prevalent, debilitating and costly diseases. For diabetes, frequent insulin treatment is very expensive and may increase anti-insulin antibody production, which may cause unwanted side effects. Corosolic acid may also have some efficacy in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service