Skip to Content
Merck
All Photos(1)

Key Documents

86852

Supelco

Tetrabutylammonium chloride

suitable for ion pair chromatography, LiChropur, ≥99.0%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[CH3(CH2)3]4NCl
CAS Number:
Molecular Weight:
277.92
Beilstein:
3571227
EC Number:
MDL number:
UNSPSC Code:
12000000
PubChem Substance ID:
NACRES:
NB.21

description

cationic

Quality Level

Assay

≥99.0% (AT)
≥99.0%

form

crystals

quality

LiChropur

technique(s)

ion pair chromatography: suitable

λ

10 % in H2O

UV absorption

λ: 220 nm Amax: 0.05
λ: 230 nm Amax: 0.04
λ: 250 nm Amax: 0.03
λ: 260 nm Amax: 0.02
λ: 500 nm Amax: 0.02

suitability

corresponds to standard for filter test

SMILES string

[Cl-].CCCC[N+](CCCC)(CCCC)CCCC

InChI

1S/C16H36N.ClH/c1-5-9-13-17(14-10-6-2,15-11-7-3)16-12-8-4;/h5-16H2,1-4H3;1H/q+1;/p-1

InChI key

NHGXDBSUJJNIRV-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Tetrabutylammonium chloride (TBACl) is a quaternary ammonium salt.g

Application

TBACl has been used as ion-pairing reagent in the mobile phase during the quantification of nitrate and nitrite in water and food samples by normal phase ion-pair high performance liquid chromatography.

Legal Information

LiChropur is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Simultaneous determination of nitrite and nitrate by normal phase ion-pair liquid chromatography.
Butt SB, Riaz M, Iqbal MZ.
Talanta, 55(4), 789-797 (2001)
Fabio Galeotti et al.
Journal of chromatography. A, 1284, 141-147 (2013-03-05)
In this study, by using tetrabutylammonium bisulfate as ion-pairing reagent, we were able to separate all the main heparin/heparan sulfate disaccharides generated by the action of heparinases along with the main Hep tetrasaccharide possessing a 3-O-sulfate group on the sulfoglucosamine
Erbo Shi et al.
Organic letters, 14(13), 3384-3387 (2012-06-27)
A metal-free C-H oxidation for the construction of allylic esters has been developed. The use of a commercially available and inexpensive catalyst and oxidant, and readily available starting materials, coupled with the operational simplicity of the reaction, renders the methodology
Morteza Moradi et al.
The Analyst, 137(15), 3549-3557 (2012-06-27)
Hollow fiber-based liquid phase microextraction (HF-LPME) using conventional solvents is limited by their relative instability and high volatility. The use of supramolecular solvents as a liquid membrane phase could overcome these inconveniences due to their negligible vapour pressure and high
Muhammet Uyanik et al.
Science (New York, N.Y.), 345(6194), 291-294 (2014-07-19)
The diverse biological activities of tocopherols and their analogs have inspired considerable interest in the development of routes for their efficient asymmetric synthesis. Here, we report that chiral ammonium hypoiodite salts catalyze highly chemo- and enantioselective oxidative cyclization of γ-(2-hydroxyphenyl)ketones

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service