Skip to Content
Merck
All Photos(1)

Key Documents

81288

Supelco

Poly(ethylene glycol)

analytical standard, for GPC, 1,000

Synonym(s):

PEG

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H(OCH2CH2)nOH
CAS Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

analytical standard
for GPC

Quality Level

shelf life

limited shelf life, expiry date on the label

mol wt

Mn ~900
Mp ~970
Mw ~940

technique(s)

gel permeation chromatography (GPC): suitable

Mw/Mn

~1.05

application(s)

cleaning products
cosmetics
food and beverages
personal care

format

neat

storage temp.

2-8°C

SMILES string

C(CO)O

InChI

1S/C2H6O2/c3-1-2-4/h3-4H,1-2H2

InChI key

LYCAIKOWRPUZTN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Poly(ethylene glycol) is a nontoxic and water-soluble polymer[1], which may find applications in drug delivery systems.[2]

Application


  • Disulfide-Cross-Linked Tetra-PEG Gels: Recent advancements in biocompatible polymers include the development of disulfide-cross-linked tetra-PEG gels, which hold significant potential in drug delivery systems due to their enhanced mechanical properties and stability (Meng et al., 2024).

  • Solvent-Free Sample Preparation for Polymer Analysis: Poly(ethylene glycol) (PEG) has been utilized in innovative solvent-free sample preparations for matrix-assisted laser desorption/ionization time-of-flight mass spectrometry, improving the analysis of polymer blends essential in biotechnological applications (Dalgic and Weidner, 2024).

  • Mammalian Cell Membrane Hybrid Polymersomes: PEG-based mammalian cell membrane hybrid polymersomes have been developed for mRNA delivery, showcasing the utility of PEG in creating biocompatible and efficient drug delivery vehicles (Shin et al., 2024).

  • Modular Synthesis of PEG-Dendritic Block Copolymers: The modular synthesis of PEG-dendritic block copolymers for drug delivery demonstrates the versatility of PEG in forming tailored nanostructures for targeted therapeutic applications (Parcero-Bouzas et al., 2024).

  • Improvement in Bioplastics Properties: Poly(ethylene glycol) has been employed to enhance the crystallization, thermal, and mechanical properties of bioplastics, highlighting its role in advancing polymer research for sustainable materials development (Pakkethati et al., 2024).

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Poly (ethylene glycol)-containing hydrogels in drug delivery
Peppas.AN, et al.
Journal of Controlled Release : Official Journal of the Controlled Release Society, 62, 81-87 (1999)
Poly (ethylene glycol) in drug delivery: pros and cons as well as potential alternatives.
Katrin K, et al.
Angewandte Chemie (International Edition in English), 49, 6288-6308 (2010)
I L Konorova et al.
Patologicheskaia fiziologiia i eksperimental'naia terapiia, (4)(4), 7-9 (1991-07-01)
The search for antiaggregatory compounds is undertaken, as a rule, under in vitro conditions which do not reflect the dynamics of the real process. The present work deals with study of the peculiarities of the development of the collagen induced
P I Polimeni et al.
Journal of cardiovascular pharmacology, 14(3), 374-380 (1989-09-01)
The acute hemodynamic effects of an intravenously (i.v.) injected poly(ethylene oxide), Polyox WSR N-60K (dose 50 mg/kg), were studied in the open-chest rat anesthetized with sodium pentobarbital. The injectate is one of four drag-reducing polymers known to augment in vitro
Mark A Rice et al.
Acta biomaterialia, 5(1), 152-161 (2008-09-17)
Ultrasound has potential as a non-destructive analytical technique to provide real-time online assessments of matrix evolution in cell-hydrogel constructs used in tissue engineering. In these studies, chondrocytes were encapsulated in poly(ethylene glycol) hydrogels, and gel degradation was manipulated to provide

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service