Skip to Content
Merck
All Photos(1)

Key Documents

69028

Supelco

4-Phenyl-α-cyanocinnamamide

matrix substance for MALDI-MS, ≥98.5% (HPLC)

Synonym(s):

4-Phenyl-α-cyanocinnamic acid amide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H12N2O
CAS Number:
Molecular Weight:
248.28
Beilstein:
3612052
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.21
Pricing and availability is not currently available.

grade

matrix substance for MALDI-MS

Quality Level

Assay

≥98.5% (HPLC)

form

powder

technique(s)

MALDI-MS: suitable

cation traces

Ba: ≤5 mg/kg
Ca: ≤20 mg/kg
Cd: ≤5 mg/kg
Co: ≤5 mg/kg
Cr: ≤5 mg/kg
Cu: ≤5 mg/kg
Fe: ≤100 mg/kg
K: ≤20 mg/kg
Mg: ≤5 mg/kg
Mn: ≤5 mg/kg
Na: ≤50 mg/kg
Ni: ≤5 mg/kg
Pb: ≤5 mg/kg
Zn: ≤5 mg/kg

SMILES string

NC(=O)\C(=C\c1ccc(cc1)c2ccccc2)\C#N

InChI

1S/C16H12N2O/c17-11-15(16(18)19)10-12-6-8-14(9-7-12)13-4-2-1-3-5-13/h1-10H,(H2,18,19)/b15-10+

InChI key

FOQBACWKDMSJBT-XNTDXEJSSA-N

related product

Product No.
Description
Pricing

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 4

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service