Skip to Content
Merck
All Photos(1)

Key Documents

45315

Supelco

Acephate

PESTANAL®, analytical standard

Synonym(s):

O,S-Dimethyl N-acetylphosphoramidothioate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H10NO3PS
CAS Number:
Molecular Weight:
183.17
Beilstein:
1936365
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

storage temp.

2-8°C

SMILES string

COP(=O)(NC(C)=O)SC

InChI

1S/C4H10NO3PS/c1-4(6)5-9(7,8-2)10-3/h1-3H3,(H,5,6,7)

InChI key

YASYVMFAVPKPKE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Mingli Wang et al.
Nanomaterials (Basel, Switzerland), 8(5) (2018-05-02)
Rapid sampling and multicomponent analysis are vital in pesticide residue detection. In this work, we proposed a SERS platform to detect three kinds of pesticides on apple peels simultaneously by a straightforward “press and peel off” method. The flexible Au/dragonfly
Nathan S Little et al.
PloS one, 14(3), e0212567-e0212567 (2019-03-14)
Interpreting variable laboratory measurements of Helicoverpa zea Boddie susceptibility to toxins from Bacillus thuringiensis Berliner (Bt) has been challenging due to a lack of clear evidence to document declining field control. Research that links laboratory measurements of susceptibility to survival
Guochao Shi et al.
Beilstein journal of nanotechnology, 10, 578-588 (2019-03-16)
In this work, we report a biomimetic synthesis route of 3D Ag nanofilm/glasswing butterfly wing hybrids (Ag-G.b.) by magnetron sputtering technology. The 3D surface-enhanced Raman scattering (SERS) substrate is fabricated from an original chitin-based nanostructure, which serves as a bio-scaffold
Lakshmipathi Srigiriraju et al.
Archives of insect biochemistry and physiology, 72(2), 105-123 (2009-06-27)
Organophosphates and carbamates represent alternative insecticides in managing the tobacco-adapted form of the green peach aphid (TGPA), Myzus persicae (Sulzer), a major pest of tobacco in the United States and around the world. General esterases that detoxify these insecticides were
Dong-Fang Hao et al.
Ecotoxicology and environmental safety, 83, 25-33 (2012-06-26)
To study the toxic effect of chronic exposure to acephate at low-dose levels, a metabolomics approach based on ultra-performance liquid chromatography/mass spectrometry (UPLC-MS) was applied. Three different doses of 0.5 mg/kg/day, 1.5 mg/kg/day, and 4.5 mg/kg/day acephate were administered to

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service