Skip to Content
Merck
All Photos(1)

Key Documents

36106

Supelco

D-α-Hydroxyglutaric acid-13C5 disodium salt

analytical standard

Synonym(s):

2-Hydroxyglutarate-13C5, (R)-2-Hydroxypentanedioic acid-13C5 disodium salt, Disodium (R)-2-hydroxyglutarate-13C5

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
13C5H6Na2O5
CAS Number:
Molecular Weight:
197.04
MDL number:
UNSPSC Code:
41116107
NACRES:
NB.21

grade

analytical standard

Quality Level

isotopic purity

≥98.0%

Assay

≥98.0% (GC)

optical activity

[α]/D 8.5±1.5°, c = 1 in 0.1 M NaOH

shelf life

limited shelf life, expiry date on the label

application(s)

clinical testing

format

neat

mass shift

M+5

storage temp.

2-8°C

SMILES string

O=[13C]([13CH2][13CH2][13C@@H](O)[13C]([O-])=O)[O-].[Na+].[Na+]

InChI

1S/C5H8O5.2Na/c6-3(5(9)10)1-2-4(7)8;;/h3,6H,1-2H2,(H,7,8)(H,9,10);;/q;2*+1/p-2/t3-;;/m1../s1/i1+1,2+1,3+1,4+1,5+1;;

InChI key

DZHFTEDSQFPDPP-JVBXKYRZSA-L

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service